Skip to Content
Merck
All Photos(1)

Key Documents

H7425

Sigma-Aldrich

Anti-FLAG®-Peroxidase antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-FLAG-HRP Polyclonal Conjugate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

Quality Level

description

buffered aqueous solution

form

lyophilized solid

technique(s)

western blot: 1:2000-1:4000 using for detection of amino-terminal FLAG-BAP fusion protein in an E. coli crude cell lysate

shipped in

dry ice

storage temp.

−20°C

General description

Anti-FLAG antibody is developed in rabbits using purified FLAG fusion protein as immunogen. Whole antiserum is purified using protein A immobilized on agarose to provide the IgG fraction of the antiserum and is conjugated to horseradish peroxidase.

ANTI-FLAG recognizes the FLAG epitope located on FLAG fusion proteins. The antibody reacts with N-terminal, N-terminal-Met, and C-terminal FLAG fusion proteins by immunoblotting. Specific staining is inhibited by the FLAG peptide (N-Asp-Tyr-Lys-Asp-AspAsp-Asp-Lys-C). Applications for the conjugate include Western blots and ELISA.

Epitope tags provide a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance, or poorly immunogenic proteins when protein specific antibodies are not available. Tagging with the FLAG peptide sequence may be done at the N-terminus, N-terminus preceded by a methionine residue, C-terminus, or at internal positions of the target protein. FLAG may also be placed in association with other tags. The small size of the FLAG tag or sequence and its high hydrophilicity tend to decrease the possibility of interference with the protein expression, proteolytic maturation, antigenicity, and function. The N-terminal FLAG peptide sequence contains a unique enterokinase cleavage site allowing it to be completely removed from the purified fusion proteins. Cleavage catalyzed by Cu2+ ions of the C-terminal FLAG peptide from a fusion protein has been reported. A sequence motif with five out of eight amino acid residues identical to the FLAG peptide is found in both rat and mouse Mg2+ dependent protein β-phosphatase, as well as in the human and bovine enzyme.

Immunogen

purified FLAG fusion protein

Application

For immunoblotting, with 1:2000 – 1:4000 as an initial suggested dilution
Browse additional application references in our FLAG® Literature FLAG® Literature portal.

Physical form

Lyophilized powder

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For Research use only. Not for drug, household, or other uses. Please consult the Safety Data Sheet for information regarding hazards and safe handling practices.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R G Chubet et al.
BioTechniques, 20(1), 136-141 (1996-01-01)
The FLAG peptide, AspTyrLysAspAspAspAspLys, has been used as an epitope tag in a variety of cell types. The modification of the cytomegalovirus (CMV) promoter containing vector, pCMV5, to create two transient expression vectors designed for secretion and intracellular expression of
D P Humphreys et al.
Protein engineering, 12(2), 179-184 (1999-04-09)
The peptide sequence (N)DKTH(C) was investigated as a site for efficient, specific cleavage of a fusion protein by cupric ions using a humanised gamma1 Fab' as a model protein. The native upper hinge (N)DKTH(C) sequence was mutated to create a
A S Robeva et al.
Biochemical pharmacology, 51(4), 545-555 (1996-02-23)
An expression plasmid for mammalian cells (CLDN10B) has been modified to add nucleotides encoding hexahistidine and the FLAG peptide (H/F) to cDNAs. The new mammalian expression plasmid has been named pDoubleTrouble (pDT). The plasmid and a recombinant baculovirus were used
K Schäfer et al.
Biochemical and biophysical research communications, 207(2), 708-714 (1995-02-15)
The FLAG peptide has been widely used as a multi-purpose tag for the identification and detection of recombinant FLAG fusion proteins. The practicability of this approach depends on specific detection of FLAG fusion proteins with no or very little cross-reactivity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service