GE28-9269-81
HiScreen™ Capto™ Adhere
Cytiva 28-9269-81
About This Item
Recommended Products
ligand
N-benzyl-N-methyl ethanolamine
description
Ion Exchanger Type (value)
packaging
pkg of 1 ea
manufacturer/tradename
Cytiva 28-9269-81
parameter
44 psi
bed size
7.7 mm × 100 mm
bed volume
4.7 mL
column I.D.
7.7 mm
matrix
highly cross-linked agarose
avg. part. size
75 (d₅₀v is the median particle size of the cumulative volume distribution)
cleaning
2-14(pH interval where the medium can be subjected to, for cleaning- or sanitization-in-place.)
working range
3-12(pH interval where the medium can be operated without significant change in function.)
suitability
suitable for bioprocess medium
Related Categories
General description
Features and Benefits
- Capto adhere is a multimodal anion exchanger for intermediate purification and polishing of monoclonal antibodies after the protein A capture step, giving the possibility to design a two-step chromatographic downstream process.
- Prepacked, ready-to-use columns for convenient process development optimization.
- 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening.
- Easily connected in series to achieve 20 cm bed height.
- Small bed volume gives fast results and minimal sample/buffer consumption.
- Reproducible results, scalable to BioProcess columns packed with the same resin using the same linear fluid velocity.
Storage and Stability
Analysis Note
Legal Information
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service