Skip to Content
Merck
All Photos(6)

Key Documents

G4626

Sigma-Aldrich

L-Glutathione oxidized disodium salt

≥98%, suitable for HPLC

Synonym(s):

GSSG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30N6Na2O12S2
CAS Number:
Molecular Weight:
656.59
Beilstein:
6265389
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

L-Glutathione oxidized disodium salt, ≥98%, powder

Quality Level

Assay

≥98%

form

powder

technique(s)

HPLC: suitable

impurities

ethanol, essentially free

color

white to off-white

storage temp.

−20°C

SMILES string

[Na+].[Na+].N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C([O-])=O)C(=O)NCC(O)=O)C(=O)NCC([O-])=O)C(O)=O

InChI

1S/C20H32N6O12S2.2Na/c21-9(19(35)36)1-3-13(27)25-11(17(33)23-5-15(29)30)7-39-40-8-12(18(34)24-6-16(31)32)26-14(28)4-2-10(22)20(37)38;;/h9-12H,1-8,21-22H2,(H,23,33)(H,24,34)(H,25,27)(H,26,28)(H,29,30)(H,31,32)(H,35,36)(H,37,38);;/q;2*+1/p-2/t9-,10-,11-,12-;;/m0../s1

InChI key

DRFILBXQKYDTFW-JIWRMXRASA-L

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

(γ-Glu-Cys-Gly) (γ-Glu-Cys-Gly) [Disulfide bridge: 2a-2b]

Application

L-Glutathione oxidized disodium salt has been used in 5,5′-dithiobis-(2-nitrobenzoic acid) (DTNB)/glutathione reductase enzyme recycling method to determine plasma or tissue glutathione levels. It has also been used in NADPH-dependent glutathione disulfide reduction assay.

Biochem/physiol Actions

Oxidized L-glutathione is frequently measured in vivo as an indicator of oxidative stress. Oxidized L-Glutathione may be converted to L-glutathione by various reducing systems.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Methods for the Determination of Plasma or Tissue Glutathione Levels
Trent E, et.al
Methods in Molecular Biology, 889, 315-324 (2012)
Dietary methionine can sustain cytosolic redox homeostasis in the mouse liver
Sofi Eriksson
Nature Communications (2015)
Oxidized forms of glutathione in peripheral blood as biomarkers of oxidative stress.
Rossi R
Clinical Chemistry, 52(7), 1406-1414 (2006)
S K Tandon et al.
Toxicology letters, 145(3), 211-217 (2003-10-29)
The influence of an antioxidant agent such as N-acetyl cysteine (NAC) or mannitol on the cadmium chelating ability of monoisoamyl 2,3-dimercaptosuccinate (MiADMS) was investigated in cadmium pre-exposed rats. This ester of 2,3-dimercaptosuccinic acid (DMSA), an accepted drug for lead poisoning
Christian Stoppe et al.
Antioxidants & redox signaling, 23(11), 865-879 (2015-08-04)
Cardiac surgery involves myocardial ischemia/reperfusion (I/R) with potentially deleterious consequences. Macrophage migration inhibitory factor (MIF) is a stress-regulating chemokine-like cytokine that protects against I/R damage, but functional links with its homolog, d-dopachrome tautomerase (MIF-2), and the circulating soluble receptor CD74

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service