G3046
GDF-15 human
Synonym(s):
Growth differentiation factor 15, MIC-1, Macrophage inhibitory cyotkine-1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
human
Quality Level
recombinant
expressed in CHO cells
Assay
>97% (visualized with silver stain, SDS-PAGE)
form
lyophilized powder
mol wt
dimer 13 kDa (based on N-terminal sequencing, starting at the first histidine of the 6X histidine tag)
technique(s)
cell culture | mammalian: suitable
impurities
endotoxin, tested
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... GDF15(9518)
General description
Growth-differentiation factor-15 (GDF-15) is a stress-responsive member of the transforming growth factor-β cytokine superfamily. It is also known as PTGF-Growth-differentiation factor-15 (GDF-15) and is a stress-responsive member of the transforming growth factor-β cytokine superfamily.2 It is also known as PTGF-β, TGF-PL or MIC-1β, TGF-PL or MIC-1.
Application
Growth-differentiation factor-15 (GDF-15) has been associated in the regulation of cell survival, proliferation, and differentiation and also in the protection from ischemia/reperfusion injury. Inflammatory and apoptotic pathways are regulated in injured tissues by GDFs during disease processes.
Biochem/physiol Actions
Although it has been classified as a divergent member of the transforming growth factor-β superfamily, GDF-15 shares structural characteristics with this superfamily. It has been identified as a biomarker for human prostate cancer. Additionally, increased GDF-15 expression has been observed in breast, pancreas and colorectal cancers. Functions of this cytokine include inhibition of TNF-α production, initiation of cartilage formation, and promotion of neuronal survival.
It has been discussed to serve as a secreted biomarker for activation of the p53 pathway in human cancer.
It has been discussed to serve as a secreted biomarker for activation of the p53 pathway in human cancer.
Physical form
Lyophilized from a 0.2 μm filtered solution in 30% acetonitrile, 0.1% trifluoroacetic acid containing 50 μg of bovine serum albumin per 1 μg of GDF-15.
Analysis Note
Under reducing conditions in SDS-PAGE, each recombinant GDF-15 monomer migrates as an ~14 kDa protein.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Renal failure and the histopathological features of myeloma kidney reversed by intensive chemotherapy and peritoneal dialysis.
British medical journal (Clinical research ed.), 294(6569), 411-412 (1987-02-14)
Quarterly journal of experimental physiology (Cambridge, England), 74(6), 813-824 (1989-11-01)
In order to examine whether an aspiration mechanism exists in the vomeronasal organ (VNO) of the domestic ram, experiments were conducted in chloralose-anaesthetized animals using acute electromanometry and electrophysiological techniques. Stimulation of the ipsilateral cervical sympathetic nerve activated a mechanism
PloS one, 10(6), e0131506-e0131506 (2015-06-27)
Although inactivating frameshift mutations in the Transforming growth factor beta receptor type 2 (TGFBR2) gene are considered as drivers of microsatellite unstable (MSI) colorectal tumorigenesis, consequential alterations of the downstream target proteome are not resolved completely. Applying a click-it chemistry
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service