Skip to Content
Merck
All Photos(1)

Key Documents

F6651

Sigma-Aldrich

Monoclonal Anti-Tumor Necrosis Factor-α–Fluorescein antibody produced in mouse

clone 6402.31, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Monoclonal Anti-Tumor Necrosis Factor-a-Carboxyfluorescein, Anti-TNF-α

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

fluorescein conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6402.31, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

flow cytometry: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... TNF(7124)

Looking for similar products? Visit Product Comparison Guide

Specificity

The antibody conjugate will identify and quantitate cells containing intracellular forms of human TNF-α.

Immunogen

recombinant human TNF-α.

Physical form

Solution in saline containing up to 0.5% bovine serum albumin and up to 0.1% sodium azide.

Preparation Note

The antibody is purified from ascites fluid and conjugated to carboxyfluorescein.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service