EP022363352
Eppendorf® Safe-Lock micro test tubes
capacity 2.0 mL, clear, pkg of 500 ea
Synonym(s):
eppendorfs
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
clear
polypropylene (push fit
)
round bottom
sterility
non-sterile
feature
RCF 30,000 × g
packaging
pkg of 500 ea
manufacturer/tradename
Eppendorf® 022363352
capacity
2.0 mL
diam.
10.5 mm
Looking for similar products? Visit Product Comparison Guide
General description
Eppendorf Safe-Lock Tubes, 2.0 mL, Eppendorf Quality, colorless, 500 tubes
- Hinged Eppendorf Safe-Lock lid prevents unintentional lid opening during incubation and storage
- g-Safe®: Depending on Safe-Lock Tube variant (see manual) exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples
- Manufactured without mold release agents, softeners or biocides – Materials which could have a negative effect on bioassays
- g-Safe®: Exceptional centrifugation stability up to 25,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples
- Precise lid sealing for minimal evaporation rates during long-term storage
- Large frosted lid and surface on the side for easy labeling
- Ensured functionality from -86 °C to 100 °C
- Autoclavable when open (121 °C, 20 min)
- Also available in purity grades of lot-certified PCR clean, Biopur® and Eppendorf Forensic DNA Grade
- Also available in variants Eppendorf Protein LoBind or Eppendorf DNA LoBind for maximum recovery of valuable samples and in amber for light-sensitive samples
Features and Benefits
Eppendorf Safe-Lock Tubes offer special protection against unintentional opening and completely eminate any danger of leaks during incubation and storage
Legal Information
Biopur is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
g-Safe is a registered trademark of Eppendorf AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service