Skip to Content
Merck
All Photos(1)

Documents

EHU158451

Sigma-Aldrich

MISSION® esiRNA

targeting human DNMT3B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCCGACAGCTCTCCAATACTCAGGTTAATGCTGAAAAATCATCCAAGACAGTTATTGCAAGAGTTTAATTTTTGAAAACTGGCTACTGCTCTGTGTTTACAGACGTGTGCAGTTGTAGGCATGTAGCTACAGGACATTTTTAAGGGCCCAGGATCGTTTTTTCCCAGGGCAAGCAGAAGAGAAAATGTTGTATATGTCTTTTACCCGGCACATTCCCCTTGCCTAAATACAAGGGCTGGAGTCTGCACGGGACCTATTAGAGTATTTTCCACAATGATGATGATTTCAGCAGGGATGACGTCATCATCACATTCAGGGCTATTTTTTCCCCCACAAACCCAAGGGCAGGGGCCACTCTTAGCTAAATCCCTCCCCGTGACTGCAATAGAACCCTCTGGGGAGC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yue Zhou et al.
American journal of translational research, 11(3), 1736-1747 (2019-04-12)
Temporomandibular joint (TMJ) arthritis causes severe debilitation and has few treatment options. Here, we found a small molecule, DNA methyltransferase 3B (Dnmt3b), as a putative therapeutic target, partially rescued osteoarthritic phenotype. Dnmt3b was detected differentially expressed in cell zones of
Hiroaki Fujimori et al.
Scientific reports, 5, 18231-18231 (2015-12-17)
A comprehensive genome-wide screen of radiosensitization targets in HeLa cells was performed using a shRNA-library/functional cluster analysis and DNMT3B was identified as a candidate target. DNMT3B RNAi increased the sensitivity of HeLa, A549 and HCT116 cells to both γ-irradiation and
Kai Xu et al.
Aging, 12(23), 23668-23683 (2020-11-23)
The role of DNA methyltransferase 3B (DNMT3B) in tumorigenesis and development has been widely recognized; however, the mechanism underlying its action remains unclear. Considering its function in de novo methylation, we aimed to investigate whether DNMT3B plays its role via
Chelsea R McCoy et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 39(16), 3144-3158 (2019-01-27)
There is growing evidence of abnormal epigenetic processes playing a role in the neurobiology of psychiatric disorders, although the precise nature of these anomalies remains largely unknown. To study neurobiological (including epigenetic) factors that influence emotionality, we use rats bred
Xin Yue et al.
Virology journal, 8, 118-118 (2011-03-16)
The hepatitis B virus (HBV) is a major etiological factor of inflammation and damage to the liver resulting in hepatocellular carcinoma. Transcription factors play important roles in the disordered gene expression and liver injury caused by HBV. However, the molecular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service