Skip to Content
Merck
All Photos(1)

Key Documents

EHU092591

Sigma-Aldrich

MISSION® esiRNA

targeting human ATF3 (1)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTTTTCTCAGATCTGGTTTCTAAGAGTTTTGGGGGGCGGGGCTGTCACCACGTGCAGTATCTCAAGATATTCAGGTGGCCAGAAGAGCTTGTCAGCAAGAGGAGGACAGAATTCTCCCAGCGTTAACACAAAATCCATGGGCAGTATGATGGCAGGTCCTCTGTTGCAAACTCAGTTCCAAAGTCACAGGAAGAAAGCAGAAAGTTCAACTTCCAAAGGGTTAGGACTCTCCACTCAATGTCTTAGGTCAGGAGTTGTGTCTAGGCTGGAAGAGCCAAAGAATATTCCATTTTCCTTTCCTTGTGGTTGAAAACCACAGTCAGTGGAGAGATGTTTGGAAACCACAGTCAGTGGAGCCTGGGTGGTACCCAGGCTTTAGCATTATTGGATGTCAATAGCA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liugen Gu et al.
Pathology, research and practice, 214(6), 862-870 (2018-05-03)
Intestinal epithelial cell (IEC) apoptosis plays a vital role in the pathogenesis of Crohn's disease (CD), which is an inflammatory bowel disease (IBD). Activating transcription factor 3 (ATF3) modulates apoptosis under stress via regulating the p53 pathway. However, the expression
Siqi Ma et al.
International journal of molecular medicine, 35(6), 1561-1573 (2015-04-16)
Glioblastomas are highly malignant gliomas that are extremely invasive with high rates of recurrence and mortality. It has been reported that activating transcription factor 3 (ATF3) is expressed in elevated levels in multiple malignant tumors. The purpose of this study
Shunyan Weng et al.
BMC gastroenterology, 16, 25-25 (2016-02-27)
Hepatocellular carcinoma (HCC) is one of most common and aggressive human malignancies in the world, especially, in eastern Asia, and its mortality is very high at any phase. We want to investigate mechanism of niclosamide inducing cell apoptosis in HCC.
Chun-Chao Chen et al.
European journal of pharmacology, 859, 172542-172542 (2019-07-19)
Nicorandil is an adenosine triphosphate-sensitive potassium channel opener with additional antioxidant properties. Doxorubicin (DOX) is an anticancer drug that exerts oxidation-mediated adverse cardiovascular effects. This study examined the effects of nicorandil on DOX-induced cytotoxicity in human umbilical vein endothelial cells
Xiaolin Liu et al.
Frontiers in physiology, 11, 540591-540591 (2021-02-05)
Mechanical stretch promotes deregulation of vascular smooth muscle cell (VSMC) functions during hypertension-induced vascular remodeling. ACE2 has a wide range of cardiovascular and renal protective effects. Loss of ACE2 is associated with cardiovascular disease, but little is known about the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service