Skip to Content

EC200

GenElute-E Single Spin Blood DNA High Yield Kit

Reagents and consumables for liquid blood samples. 10, 50 or 250 purifications.

greener alternative

Greener Alternative

Synonym(s):

GenElute-E Blood DNA High Yield Kit, GenElute-E kits

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.52
UNSPSC Code:
41105600
Technique(s):
DNA purification: suitable
Storage temp.:
2-8°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


purified by

(Single-spin negative chromotography), (Time: 30 minutes or less)

feature

Compatible Application (Suitable for most common downstream applications, including genotyping, PCR, and NGS), Intended use (For the purification of genomic DNA from liquid blood samples), Typical/expected yield (Varies by sample. Please reference user guide for more information.)

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

DNA purification: suitable

input

blood (liquid or dried)

test parameters

: 3 min hands on time, sample volume: 200-1000 μL blood

greener alternative category

storage temp.

2-8°C

General description

GenElute-E purification kits provide a simple alternative for DNA purification. Compared with traditional methods, GenElute-E offers:
  • Isolation of DNA in under an hour with minimal sample handling
  • Better purity, leading to improved performance in PCR and other applications
  • Significant reduction in plastic and hazardous chemical waste


Traditional silica-based, bind-wash-elute purification kits require multiple wash steps to remove impurities from the spin columns. These steps increase the risk of cross contamination, subject the DNA to centrifugation sheering forces, and introduce chaotropic salts that can carry over into the final sample, inhibiting downstream applications.

GenElute-E kits employ size exclusion negative chromatography to separate large nucleic acid molecules from smaller protein, lipid, and ionic components in cell, tissue, blood, and other samples. Single-spin columns efficiently absorb and retain cellular debris and sample contaminants while allowing nucleic acids to pass through, reducing the number of steps and plastic materials required for purification. This novel method for high-quality purification is made possible by our innovative SmartLyse® Protease, which enables fast and efficient lysis of a wide range of sample types.

Isolate nucleic acid in a fraction of time compared to traditional silica bind-wash-elute procedures. Simply mix sample with lysis components and incubate according to kit protocol, pipette sample onto the spin column, and centrifuge directly into a collection tube. Cellular debris and contaminants remain bound in the column to be discarded, while purified DNA is ready to be used in downstream applications or stored.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is aligned with "Waste prevention" and "Designing safer chemicals". Click here for more information.

Application

Store enzymes at 2-8°C for up to 12 months. All other components should be stored at room temperature.
Traditional methods for purifying genomic DNA from liquid blood samples ≥ 200 μl rely on medium and large format silica (bind-wash-elute) purification kits, which suffer from limited binding capacity, low DNA concentration, and low purity. The GenElute-E Single Spin Blood DNA High Yield Kit, extracts up to 20 μg highly concentrated DNA (200 ng/μl) while offering the flexibility of sample inputs ranging from 200 μl to 1 ml liquid blood. EDTA-, citrate- as well as heparin-stabilized human and animal blood can be used. Enriched DNA yield and concentration make this kit well suited for researchers with limited or low-titer samples who require high sensitivity for NGS or other detection methods.Purified genomic DNA is eluted in Tris buffer, pH 7.8 and can immediately be used for most common downstream applications, including PCR, genotyping, NGS, and others. Genomic DNA preparations usually show an A260/280 ratio of 1.8 to 2.0.

Features and Benefits

Fast and efficient purification: 10-minute lysis and minimal hands-on, preparation stepsHigher quality DNA: negative chromotography-based purification reduces carryover of PCR inhibitors and other contaminents Reduced environmental impact: 55% less plastic waste, as well as a significant reduction in hazardous chemicals

Other Notes

We believe that green chemistry will contribute to a better tomorrow. With a growing portfolio of greener alternatives, there are now more choices to reduce the ecological impact of your research while still delivering quality and efficacy so your results are not compromised.  GenElute-E purification kits avoid the use of hazardous wash buffers typically employed in silica-based purification kits, significantly reducing chemical waste. The single-spin protocol also limits the number of collection steps, reducing plastic waste by 55% compared to equivalent bind-wash-elute methods. GenElute-E is designated a Greener Alternative Product based on the "Prevention" and "Designing Safer Chemicals" qualifications of The 12 Principles of Green Chemistry.GenElute-E kits also adhere to the principles of SMASH Packaging, our global initiative to improve the sustainability through the reduction of packaging materials, the increased use of sustainable alternatives, and improved recyclability. Packaging is FCS-marked for sustainable forestry practices and produced from greater than 70% recycled content.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC
SmartLyse is a registered trademark of Merck KGaA, Darmstadt, Germany


Kit Components Only

Product No.
Description

  • Erythrocyte Lysis Buffer

  • Blood Lysis Buffer HY

  • SmartLyse® HY Protease

  • Clearing Solution HY

  • 1x Tris Buffer

  • Spin Columns

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Resp. Sens. 1

Storage Class

3 - Flammable liquids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library