Skip to Content
Merck

D6315

Nunclon® Δ Multidishes

12 wells, flat bottom

Synonym(s):

12 well plate, multiwell plate, Nunc 150628, cell cultivation plate, culture plate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.22
UNSPSC Code:
41122107
Material:
flat bottom clear polystyrene wells
Size:
12 wells
Sterility:
sterile; γ-irradiated
Binding type:
Tissue Culture (TC)-treated surface
Feature:
lid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

flat bottom clear polystyrene wells

sterility

sterile; γ-irradiated

feature

lid

packaging

case of 75 ea (individually wrapped)

manufacturer/tradename

Nunc 150628

maximum volume

2 mL

size

12 wells

well maximum volume

2 μm

working volume

2 mL

binding type

Tissue Culture (TC)-treated surface

General description

Nunclon® Delta multidishes are useful in all areas of cell culture e.g. scale-up and cloning. SonicSeal slide wells for cell culture in situ on Permanox® slide. Permanox is not auto-fluorescent. Raised well rims to lower risk of cross contamination. Excellent optical quality. Certified surface treatment ensures optimal condition for cell attachment and growth.

Features and Benefits

Nunclon®Δ Multidishes with Round Well Styles feature:
  • Available as flat well bottom and stackable
  • Sterilized using gamma irradiation
  • Certified as non-pyrogenic
  • Proper identification of well using alphanumerical codes
  • Maximum adhesion ensured by proprietary surface modification
  • Vented lids to enable gas exchange
  • Recessed areas around wells act as a reservoir to reduce evaporation from wells
  • Corner notch design of the Lids allow easy orientation and reduce cross-contamination

Legal Information

Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Permanox is a registered trademark of Thermo Fisher Scientific or its subsidiaries


Still not finding the right product?

Explore all of our products under Nunclon® Δ Multidishes




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library