CLS4872
Corning® Costar® reagent reservoirs
white polystyrene, capacity 100 mL, pack of 200 ea (Bulk)
Synonym(s):
Pipette tips for Velocity
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121522
NACRES:
NB.14
Recommended Products
material
U-bottom bottom
white polystyrene
sterility
sterile
packaging
pack of 200 ea (Bulk)
manufacturer/tradename
Corning 4872
capacity
100 mL
Looking for similar products? Visit Product Comparison Guide
General description
Corning® Costar® reagent disposable reservoirs are used to repetitively fill multichannel pipettors. These reserviors are non-pyrogenic and free of DNase/RNase contamination.
Application
Corning® Costar® reagent reservoirs haVE been used with multi-channel pipettors for seamless transfer of samples, buffers, or reagents into ELISA microplates.
Features and Benefits
- Sterile, disposable reservoirs for efficient reagent handling
- Non-pyrogenic
- Free from DNase/RNase contamination
- Made from modified polystyrene (white)
- Convenient 100 mL capacity
- Compatible with multichannel pipettors
Legal Information
Corning is a registered trademark of Corning, Inc.
Costar is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service