Skip to Content
Merck
All Photos(1)

Key Documents

CLS3990

Corning® 96 well NBS Microplate

flat bottom, white polystyrene, bag of 5 ×, non-sterile, lid: no

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.15
material:
flat bottom
polystyrene
white bottom
white polystyrene
maximum volume:
360 μL
size:
96 wells
well volume:
360 μL
well working volume:
75-200 μL
working volume:
200 μL
Pricing and availability is not currently available.

material

flat bottom
polystyrene
white bottom
white polystyrene

sterility

non-sterile
non-sterile

feature

lid: no
lid: no
skirt
plate format: 96 well standard

packaging

case of 100
pack of 25
bag of 5 ×

manufacturer/tradename

Corning 3990

maximum volume

360 μL

size

96 wells

well volume

360 μL

well working volume

75-200 μL

working volume

200 μL

color

white

binding type

NBS (non-binding surface)

Looking for similar products? Visit Product Comparison Guide

General description

Non-Binding surface (NBS) multiwell plates have a modified polymer surface (PS) which results in a non-ionic hydrophilic surface (polyethylene oxide-like), thereby minimizing bio-interaction and enhancing the signal-to-noise ratio. Results have demonstrated NBS′s ability to significantly reduce protein and nucleic acid binding to polymers (<2 ng/cm2; over 90% reduction of protein binding compare to PS & PP surface), maintain enzyme activity, and inhibit adhesion of a number of cell lines. These novel surface technologies for assay plates are ideal for homogeneous assays such as Scintillation Proximity Assay (SPA). In addition to a 20+ fold increase in signal-to-noise ratio, selected assays have also displayed improved sensitivity and specificity. The recommended working volume of up to 75 μL to 200 μL for 96 well plates and 20μL to 80 μL for 384 well plates.

Legal Information

Corning is a registered trademark of Corning, Inc.
NBS is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdisalan M Noor et al.
BMC infectious diseases, 11, 121-121 (2011-05-17)
Countries aiming for malaria elimination require a detailed understanding of the current intensity of malaria transmission within their national borders. National household sample surveys are now being used to define infection prevalence but these are less efficient in areas of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service