Skip to Content
Merck
All Photos(1)

Key Documents

CLS3897

Corning® 96 Well Clear Polystyrene Microplate

V-bottom clear, polystyrene, bag of 25, non-sterile, lid: no

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
V-bottom clear
polystyrene
round wells
size:
96 wells
well volume:
320 μL
well working volume:
75-200 μL
Pricing and availability is not currently available.

material

V-bottom clear
polystyrene
round wells

sterility

non-sterile

feature

lid: no
skirt

packaging

bag of 25

manufacturer/tradename

Corning 3897

size

96 wells

well volume

320 μL

well working volume

75-200 μL

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Corning® 96 well clear microplates support various application requirements.
  • Tissue culture-treated (TC-treated) polystyrene surface is used for the attachment and growth of anchorage-dependent cells.
  • V Bottom is best suited for retrieving - with least dead volume.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdisalan M Noor et al.
BMC infectious diseases, 11, 121-121 (2011-05-17)
Countries aiming for malaria elimination require a detailed understanding of the current intensity of malaria transmission within their national borders. National household sample surveys are now being used to define infection prevalence but these are less efficient in areas of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service