Key Documents
CLS3303
Corning® CellSTACK® cell culture chambers
polystyrene, sterile (γ-irradiated), 1 Chamber with Vent Caps, graduations, case of 8
Synonym(s):
cell culture flasks, scale up cell culture flasks, tissue culture flasks
About This Item
UNSPSC Code:
41122104
NACRES:
NB.13
Pricing and availability is not currently available.
material
cap (vented)
polystyrene
rectangular
rectangular bottom flask
straight neck
vent cap
sterility
sterile (γ-irradiated)
feature
cap
graduations
packaging
pack of 1
case of 8
manufacturer/tradename
Corning 3303
surface area
636 cm2
working volume
125-190 mL
suitability
suitable for (scale-up cell culture)
binding type
low binding surface
General description
  • Ideal for cell culture scale-up
  • Culture chambers produce cell yields in the 107 - 109 range
  • Useful alternatives to multiple roller bottles or spinner flasks.
  • High mechanical strength and structural integrity
  • 100% leak tested prior to shipping
  • Certified nonpyrogenic and sterilized by gamma irradiation, with low particulate contamination from modern manufacturing methods
  • Easy to use larger openings with threaded closures and vented caps
  • Large 26 mm wide filling ports allow direct access to chamber bottom, with more options for sterile transfer of media and cultures
Features and Benefits
  • Cell growth area is 636 cm2.
  • Ultra Low Attachment surface features a covalently bound hydrogel layer that effectively inhibits cellular attachment and minimizes protein absorption, enzyme activation, and cellular activation.
  • Surface is non-cytotoxic, biologically inert, and nondegradable.
  • Two 26 mm diameter filling allow direct access to chamber bottom providing greater flexibility for sterile filling and emptying by pouring, pipetting or via tubing in a fully closed system.
  • Standard 33 mm threaded orange caps have 0.2 μm pore nonwettable membranes sealed directly to the caps to allow gas exchange (vented caps) while minimizing the risk of contamination.
  • Optional 33 mm threaded caps are available with integrally sealed USP Class VI certified C-Flex tubing to allow direct sterile transfer of media and cells.
  • USP Class VI polystyrene provides excellent optical clarity and mechanical strength.
  • Sterilized by gamma radiation and certified nonpyrogenic.
  • 100% leak tested prior to shipping.
Legal Information
CellSTACK is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)