Skip to Content
Merck
All Photos(1)

Key Documents

F-36P

99072808, human bone, Lymphoblastoid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
human bone
growth mode:
Suspension
karyotype:
43,Y(Xp+,-5,-7,-13,-16,-17,-19,-21,2q-,9p+,10q+,+4mar)
morphology:
Lymphoblastoid
products:
Not specified
receptors:
Not specified
relevant disease(s):
cancer

Product Name

F-36P, 99072808

biological source

human bone

growth mode

Suspension

karyotype

43,Y(Xp+,-5,-7,-13,-16,-17,-19,-21,2q-,9p+,10q+,+4mar)

morphology

Lymphoblastoid

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human leukemia, myelodysplastic syndrome

Cell Line Description

The cell line F-36P has been established from a 65 year old male patient, who was diagnosed with refractory anaemia with excess blasts (RAEB), a subtype of myelodysplastic syndrome (MDS). This non-lymphoid leukemia line was obtained from the patient′s pleural fluid at time of progressed to overt leukemia. F-36P is cytokine-dependent requiring granulocyte-macrophage colony-stimulating factor (GM-CSF) or Interleukin 3 (IL-3). Cells have been shown to synthesize haemoglobin when GM-CSF or IL-3 is substituted by erythropoietin (Epo). They have been reported to be positive for leukocyte common antigen (CD45) and some multilineage markers such as CD13, CD33 and CD34, but are negative for T- and B-cell antigens and mature myelomonocytic antigens. Due to their reaction with some monoclonal antibodies recognising erythroid and platelet glycoproteins it has been suggested that F-36P has a multilineage phenotype.

Application

Study of differentiation and effect of growth signals

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 9
D13S317: 10
D16S539: 10,12
D5S818: 13
D7S820: 9,11
THO1: 8
TPOX: 9,11
vWA: 14,16

Culture Medium

RPMI 1640 + 2mM Glutamine + 5ng/ml rh IL-3 + 5% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9 x 100,000 cells/ml; 5%CO2; 37ºC.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service