Skip to Content
Merck
All Photos(1)

Documents

SR-4987

94102704, mouse bone, Fibroblast-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

SR-4987, 94102704

biological source

mouse bone

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast-like

products

Macrophage colony stimulating factor (M-MCF)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse bone marrow stromal cells

Cell Line Description

A bone marrow-derived stromal cell line established by A Pessina in 1987 from a normal female 5 month old mouse. A bone marrow culture was treated with the supernatant from Y1 cells which actively produce viral C-particles (MuLV). This fibroblast-like cell line produces macrophage-colony stimulating factor (M-MCF) and exhibits a high sensitivity to fibroblast growth factor (FGF) in clonogenic assays. The cells produce sarcomas in syngenic mice and have a reported doubling time of 14.7 hours.

Culture Medium

McCoy′s 5a + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:8 i.e. seeding at 1-2x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service