Skip to Content
Merck
All Photos(1)

Documents

G

94101453, rat prostate, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

G, 94101453

biological source

rat prostate

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Androgen sensitive

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat Dunning R-3327,prostate adenocarcinoma

Cell Line Description

Derived from a Dunning R-3327 rat with prostatic adenocarcinoma. G is one of a group of variants which developed by a single in vivo passage. It is androgen responsive, moderately fast growing and poorly differentiated. Using this system of rat prostatic adenocarcinomas as a model, it has been demonstrated that prostatic cancer cells can become unstable at any time resulting in the addition of cytogenetically and phenotypically novel clones of cancer cells to the cancer population.

Culture Medium

RPMI 1640 + 2mM Glutamine + 250nM Dexamethasone (DXMT) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:10 i.e. seeding at 1-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service