Skip to Content
Merck
All Photos(1)

Documents

NCI-H727

94060303, human lung, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

NCI-H727, 94060303

biological source

human lung

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Synthesise the peptide neuromedin B (NMB) (0.1 pmol/mg protein).

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human lung non-small cell carcinoma

Cell Line Description

A non-small cell carcinoma of the lung from a 65 year old Caucasian female taken, prior to treatment. The cells express easily detectable levels of p53 mRNA compared to levels found in normal lung. The cells are not able to synthesise gastrin releasing peptide (GRP). NCI-H727 is the best differentiated available bronchial carcinoid cell line. Any proposed commercial use of these cells or products must first be negotiated with the University of Texas. More details available upon request.

Application

Synthesis of NMB.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11,12
D13S317: 11
D16S539: 11,13
D5S818: 11,12
D7S820: 8,10
THO1: 8
TPOX: 8
vWA: 14,15

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service