Skip to Content
Merck
All Photos(1)

Key Documents

XrS6

92052128, hamster ovary, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
hamster ovary
growth mode:
Adherent
karyotype:
Not specified
morphology:
Epithelial
products:
Not specified
receptors:
Not specified

Product Name

XrS6, 92052128

biological source

hamster ovary

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Hamster Chinese ovary

Cell Line Description

Derived from the parent line CHO-K1 and is hypersensitive to ionising radiation but not to UV irradiation. Cells may revert to wild-type after 1-2 weeks in culture. They should be checked for sensitivity and recloned when necessary.

Application

Radiation sensitivity and mutation studies.

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:5 to 1:10 i.e. seeding at 1-2x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service