Nthy-ori 3-1 Cell Line human
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 90011609, human thyroid (follicular epithelial), Epithelial
Synonym(s):
HTori-3.1 Cells, Nthy-ori 3.1 Cells
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
biological source
human thyroid (follicular epithelial)
packaging
tube of 5 μg 90011609-DNA-5UG
pkg of vial of cells 90011609-1VL
growth mode
Adherent
karyotype
Not specified
morphology
Epithelial
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human thyroid follicular epithelial
Cell Line Description
Normal human primary thyroid follicular epithelial cells were transfected with a plasmid containing an origin-defective SV40 genome (SV-ori) to immortalise them. SV40 DNA was able to immortalise normal thyroid epithelial cells with the retention of several different functions.
Application
Nthy-ori 3-1 human cell Line has been used for transfection and virus studies. It is also used for the study of control and growth in human thyroid. The cell line has been used for microRNA (miRNA) expression profiling.
Culture Medium
RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service