Skip to Content
Merck
All Photos(1)

Key Documents

59M

89081802, human ovary, Epithelial

Synonym(s):

OAW 59M Cells, OAW59M Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

59M, 89081802

biological source

human ovary

packaging

tube of 5 μg 89081802-DNA-5UG
pkg of vial of cells 89081802-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human ovarian tumour epithelial

Cell Line Description

Derived from a patient with carcinoma of the ovary and is of ascitic fluid origin.

Application

Tumourigenicity studies

Culture Medium

DMEM + 2mM Glutamine + 1mM Sodium Pyruvate (NaP) + 20 IU/l Bovine Insulin + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 i.e. seeding at 4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.
Depositor and originator: Dr A Wilson, Oncology Research Laboratory, Derby City Hospital

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service