Skip to Content
Merck
All Photos(1)

Key Documents

KATO-III

86093004, human stomach (gastric carcinoma), Spherical

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

KATO-III, 86093004

biological source

human stomach (gastric carcinoma)

growth mode

Suspension

karyotype

2n = 46, hypertriploid

morphology

Spherical

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human gastric carcinoma

Cell Line Description

Derived from a metastasis of a gastric carcinoma in a 55-year-old Down Syndrome male.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 7,11
D13S317: 8,12
D16S539: 10,12
D5S818: 10,11
D7S820: 8,12
THO1: 7,9
TPOX: 11
vWA: 14,16

Culture Medium

RPMI 1640 + 2mM Glutamine + 20% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 2-9x100,000 cells/ml; 5% CO2; 37°C. Cells grow in suspension, but will increasingly to adhere to the surface during longer culture. For passaging it is recommended to subculture both cell types using scrapers for dislodging adh

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service