Skip to Content
Merck
All Photos(2)

Key Documents

C6805

Sigma-Aldrich

Monoclonal Anti-Collagen, Type VII antibody produced in mouse

clone LH7.2, ascites fluid

Synonym(s):

Anti-EBD1, Anti-EBDCT, Anti-EBR1, Anti-NDNC8

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

LH7.2, monoclonal

contains

15 mM sodium azide

species reactivity

human, goat, marmoset, monkey, guinea pig, sheep, pig, bovine

technique(s)

dot blot: suitable
immunocytochemistry: suitable
indirect ELISA: suitable
indirect immunofluorescence: 1:1,000 using human or other mammalian frozen sections
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COL7A1(1294)

Related Categories

General description

Collagen is a fibrous protein present in the extracellular framework of all vertebrates. Collagen type VII is the crucial component of anchoring fibrils and mutation in type VII collagen gene (COL7A1) leads to epidermolysis bullosa dystrophica. Monoclonal anti-collagen, type VII antibody can be used to distinguish invasive melanoma from non-invasive melanoma by visualizing the appearance and integrity of epidermal basement membrane. It can also be used in immunoblotting. Mouse anti-collagen type VII antibody reacts specifically with an epitope located on collagenase digested type VII collagen (150 kDa). The product also reacts with the basement membrane zone of stratified squamous epithelia of sheep, pig, bovine, guinea pig, goat and human.
Collagen type VII alpha 1 chain (COL7A1) is majorly expressed in skin, secreted by keratinocytes and fibroblasts. The gene is located on human chromosome 3p21.31.

Immunogen

insoluble fractions of neonatal human foreskin epidermal cells

Application

Monoclonal Anti-Collagen, Type VII antibody produced in mouse has been used in immuno-gold labelling.
Monoclonal anti-collagen type VII antibody can be used as a primary antibody in virus titration. It can also be used for the localization of type VII collagen in different immunochemical assays like dot blot, immunocytochemistry and ELISA.

Biochem/physiol Actions

Collagen type VII alpha 1 chain (COL7A1) is required for epithelium-to-stroma anchorage in skin, mucosa and cornea. It is considered as an immediate-early response gene for transforming growth factor β (TGF-β)/SMAD (mothers against decapentaplegic) signaling pathway.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Type VII collagen associated with the basement membrane of amniotic epithelium forms giant anchoring rivets which penetrate a massive lamina reticularis
Ockleford CD, et al.
Placenta, 34(9), 727-737 (2013)
Molecular basis of dystrophic epidermolysis bullosa: mutations in the type VII collagen gene (COL7A1).
Jarvikallio A, Pulkkinen L
Human Mutation, 10(5), 338-347 (1997)
Type VII Collagen in the Human Accommodation System: Expression in Ciliary Body, Zonules, and Lens Capsule
Wullink B, et al.
Investigative Ophthalmology & Visual Science, 59(2), 1075-1083 (2018)
R Schucht et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 14(2), 285-292 (2006-05-16)
We developed a new strategy that provides well-defined high-titer producer cells for recombinant retroviruses in a minimum amount of time. The strategy involves the targeted integration of the retroviral vector into a chromosomal locus with favorable properties. For proof of
Daisuke Sawamura et al.
The Journal of investigative dermatology, 118(6), 967-971 (2002-06-13)
The expression of intradermally injected DNA by keratinocytes is found mainly in the upper and middle layers of the epidermis. To investigate the mechanism of this selective expression, we observed the sequential changes in the distribution of interleukin-6-expressing keratinocytes after

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service