Skip to Content
Merck
All Photos(1)

Key Documents

AXYSCT15ML25S

Centrifuge tube with screw cap, conical bottom, co-polymer

capacity 15 mL, sterile

Synonym(s):

Conical centrifuge tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703
NACRES:
NB.11

material

conical bottom tube
polypropylene
screw top cap

sterility

sterile

packaging

pack of 25 ea
case of 500 ea

manufacturer/tradename

Axygen Inc SCT-15ML-25-S
Corning

parameter

12000 × g max. RCF

capacity

15 mL

color

clear

fitting

cap (screw cap)

suitability

suitable for (centrifugation)

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service