Skip to Content
Merck
All Photos(1)

Key Documents

AXYPDW10ML24C

Deep well plate

with rectangular well, size 24 wells, clear, size 10 mL, pkg of 5x5plates/cs, non-sterile

Synonym(s):

Deep Well Plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
material:
clear bottom
rectangular bottom wells
size:
10 mL
24 wells

material

clear bottom
rectangular bottom wells

sterility

non-sterile

feature

lid: no
skirt

manufacturer/tradename

Axygen Inc P-DW-10ML-24-C
Corning

packaging

pkg of 5x5plates/cs

size

10 mL
24 wells

color

clear

suitability

suitable for PCR

Looking for similar products? Visit Product Comparison Guide

General description

  • Axygen® deep well and assay microplates are made of chemically resistant and temperature resistant high-quality polypropylene.
  • Ideal for sample collection, invitro growth chamber, and long-term storage.
  • The round-well design of microplate ensures complete sample recovery.
  • The plates can be sealed using a heat-sealing film for sample protection.

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service