AXYAM96PCRRD
Sealing mat
for PCR microplates, size 96 wells, pkg of 5x10mats/cs
Synonym(s):
96 multiwell plate, 96 well PCR plate, 96 well microplate, 96 well microtiter plate
All Photos(1)
About This Item
UNSPSC Code:
41122410
NACRES:
NB.22
Recommended Products
material
silicone (research-grade)
sterility
non-sterile
manufacturer/tradename
Axygen Inc AM-96-PCR-RD
Corning
packaging
pkg of 5x10mats/cs
size
96 wells
Related Categories
General description
- Axygen® AxyMats® 96 Round Well Sealing Mats are ideal for a wide range of storage and PCR applications.
- Autoclavable mats are made of research-grade silicone.
- Offers ultimate protection for storage applications involving more aggressive solvents.
- The mats provide a tight seal that prevents evaporation of the samples.
- Reusable - following a bleach wash and ethanol rinse protocol.
Legal Information
AxyMats is a registered trademark of Corning, Inc.
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)