Skip to Content
Merck
All Photos(1)

Key Documents

AV49523

Sigma-Aldrich

Anti-IL22 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-IL-21, Anti-IL-22, Anti-IL-D110, Anti-IL-TIF, Anti-IL21, Anti-ILTIF, Anti-Interleukin 22, Anti-MGC79382

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

20 kDa

species reactivity

rat, mouse, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL22(50616)

General description

Interleukin 22 (IL-22) belongs to the IL10 family of T-cell associated cytokines, highly expressed in αβ and γδ T cells, as well as in innate lymphoid cells. Inflammatory cells associated with the thymus, pancreas, synovium, skin, and gut secrete IL-22. The Il-22 gene is localized on human chromosome 12q15.

Immunogen

Synthetic peptide directed towards the C terminal region of human IL22

Application

Anti-IL22 antibody produced in rabbit has been used in immunoblotting (1:1000).

Biochem/physiol Actions

Interleukin 22 (IL-22) plays a key role in cell proliferation, cellular defense, and tissue regeneration, It shows potential in wound healing and to treat gastrointestinal (GI)-related illnesses. Higher levels of IL-22 is observed in systemic lupus erythematosus, vitiligo, multiple sclerosis, etc.
Interleukin 22 (IL22) belongs to the interleukin 10 (IL-10) family. It is a cytokine that contributes to the inflammatory response in vivo. IL22 cellular effects are mediated by a heterodimeric receptor made up of IL22 and IL-10Rβ. IL22 signaling mediates proliferation of epithelial cells during inflammation mainly by the activation of signal transducer and activator of transcription 1 (STAT1) and STAT3 signaling.

Sequence

Synthetic peptide located within the following region: CHIEGDDLHIQRNVQKLKDTVKKLGESGEIKAIGELDLLFMSLRNACI

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lauren A Zenewicz
ImmunoHorizons, 2(6), 198-207 (2019-04-26)
IL-22 is a critical cytokine in modulating tissue responses during inflammation. IL-22 is upregulated in many chronic inflammatory diseases, making IL-22 biology a potentially rewarding therapeutic target. However, this is complicated by the dual-natured role of IL-22 in inflammation, as
Olivia B Parks et al.
Frontiers in cell and developmental biology, 3, 85-85 (2016-01-23)
Interleukin (IL)-22 is a member of the IL-10 family of cytokines that has been extensively studied since its discovery in 2000. This review article aims to describe the cellular sources and signaling pathways of this cytokine as well as the
Jane A Lindborg et al.
Cell reports, 34(9), 108777-108777 (2021-03-04)
Adult mammalian central nervous system (CNS) trauma interrupts neural networks and, because axonal regeneration is minimal, neurological deficits persist. Repair via axonal growth is limited by extracellular inhibitors and cell-autonomous factors. Based on results from a screen in vitro, we evaluate
Kerstin Wolk et al.
European journal of immunology, 36(5), 1309-1323 (2006-04-19)
IL-22 is an IFN-IL-10 cytokine family member, which is produced by activated Th1 and NK cells and acts primarily on epithelial cells. Here we demonstrate that IL-22, in contrast to its relative IFN-gamma, regulates the expression of only a few
Lauren A Zenewicz et al.
European journal of immunology, 38(12), 3265-3268 (2008-11-20)
IL-22 is a Th17 T-cell-associated cytokine that is highly expressed during chronic inflammation. IL-22 receptor expression is absent on immune cells, but is instead restricted to the tissues, providing signaling directionality from the immune system to the tissues. Through Stat3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service