Skip to Content
Merck
All Photos(1)

Key Documents

AV35609

Sigma-Aldrich

Anti-TRPM3 (AB3) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-GON-2, Anti-LTRPC3, Anti-MLSN2, Anti-Transient receptor potential cation channel, subfamily M, member 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
dog, mouse, human, guinea pig, bovine, rat, rabbit, horse
citations:
1
technique(s):
western blot: suitable

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

mol wt

188 kDa

species reactivity

dog, mouse, human, guinea pig, bovine, rat, rabbit, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRPM3(80036)

Immunogen

Synthetic peptide directed towards the N terminal region of human TRPM3

Biochem/physiol Actions

Transient receptor potential cation channel, subfamily M, member 3 (TRPM3) belongs to the TRP family of channels. These cation-selective channels that regulate homeostasis, calcium signaling, calcium entry and storage.

Sequence

Synthetic peptide located within the following region: ALVACKLCKAMAHEASENDMVDDISQELNHNSRDFGQLAVELLDQSYKQD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Oberwinkler et al.
Handbook of experimental pharmacology, (179)(179), 253-267 (2007-01-16)
TRPM3 is the last identified member of the TRPM subfamily and is most closely related to TRPM1. Due to alternative splicing, the TRPM3 gene encodes a large number of different variants. One splice event, affecting the pore-forming region of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service