Skip to Content
Merck

96317

Sigma-Aldrich

6-Chloro-3-indolyl α-D-galactopyranoside

≥98.0% (HPLC), powder

Synonym(s):

Salmon α-Gal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16ClNO6
CAS Number:
Molecular Weight:
329.73
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Pricing and availability is not currently available.

product name

6-Chloro-3-indolyl α-D-galactopyranoside, ≥98.0% (HPLC)

Assay

≥98.0% (HPLC)

form

powder

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@H](Oc2c[nH]c3cc(Cl)ccc23)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C14H16ClNO6/c15-6-1-2-7-8(3-6)16-4-9(7)21-14-13(20)12(19)11(18)10(5-17)22-14/h1-4,10-14,16-20H,5H2/t10-,11+,12+,13-,14+/m1/s1

InChI key

OQWBAXBVBGNSPW-HTOAHKCRSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
156353C69802D73405
form

liquid

form

-

form

liquid

form

crystals

bp

148 °C (lit.)

bp

-

bp

166 °C/714 mmHg (lit.)

bp

-

density

1.194 g/mL at 25 °C (lit.)

density

-

density

1.2 g/mL at 25 °C (lit.)

density

-

refractive index

n20/D 1.533 (lit.)

refractive index

-

refractive index

n20/D 1.532 (lit.)

refractive index

-

product line

ReagentPlus®

product line

-

product line

-

product line

-

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Need A Sample COA?

This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service