Skip to Content
Merck
All Photos(1)

Key Documents

55328

Sigma-Aldrich

8-Hydroxy-N,N,N′,N′,N″,N″-hexamethylpyrene-1,3,6-trisulfonamide

suitable for fluorescence, ≥95% (HPCE)

Synonym(s):

8-Hydroxypyrene-1,3,6-tris(dimethylsulfonamide)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H25N3O7S3
CAS Number:
Molecular Weight:
539.64
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32

Assay

≥95% (HPCE)

form

solid

pKa 

5.7

mp

276 °C (lit.)

solubility

DMF: soluble
DMSO: soluble
alcohols: soluble

fluorescence

λex 490 nm; λem 545 nm in 0.1 M Tris pH 9.0

suitability

suitable for fluorescence

SMILES string

CN(C)S(=O)(=O)c1cc(O)c2ccc3c(cc(c4ccc1c2c34)S(=O)(=O)N(C)C)S(=O)(=O)N(C)C

InChI

1S/C22H25N3O7S3/c1-23(2)33(27,28)18-11-17(26)13-7-8-15-19(34(29,30)24(3)4)12-20(35(31,32)25(5)6)16-10-9-14(18)21(13)22(15)16/h7-12,26H,1-6H3

InChI key

IMRMTOQIIAICNM-UHFFFAOYSA-N

Application

suitable as pH-indicator

Other Notes

Fluorescent pH indicator for the weakly acidic pH range and enzyme substrate reference standard

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

E. Koller et al.
Appl. Fluoresc. Technol., 1, 15-15 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service