Skip to Content
Merck
All Photos(3)

Key Documents

484R-1

Sigma-Aldrich

PRAME (EP461) Rabbit Monoclonal Primary Antibody

Synonym(s):

Preferentially-expressed Antigen in Melanoma

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP461, monoclonal

description

For In Vitro Diagnostic Use in Select Regions

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (484R-14)
vial of 0.1 mL concentrate Research Use Only (484R-14-RUO)
vial of 0.5 mL concentrate (484R-15)
vial of 1.0 mL concentrate (484R-16)
vial of 1.0 mL concentrate Research Use Only (484R-16-RUO)
vial of 1.0 mL pre-dilute Research Use Only (484R-17-RUO)
vial of 1.0 mL pre-dilute ready-to-use (484R-17)
vial of 25.0 mL pre-dilute ready-to-use Research Use Only (484R-10-RUO)
vial of 25.0 mL pre-dilute ready-to-use (484R-10)
vial of 7.0 mL pre-dilute ready-to-use (484R-18)
vial of 7.0 mL pre-dilute ready-to-use Research Use Only (484R-18-RUO)

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:50 (concentrated)

isotype

IgG

control

melanoma

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

General description

PRAME (PReferentially-expressed Antigen in MElanoma) is a gene encoded on the 22q11-22 chromosomal sequence and encodes a 509 amino acid residue protein.1 PRAME is a melanoma antigen that is preferentially expressed in tumors and is recognized by cytotoxic T lymphocytes.2,3 PRAME can be used to distinguish between malignant melanoma cells and nevus cells,4 and therefore may be useful for diagnostic purposes to support a suspected case of melanoma. PRAME is considered a cancer-testis antigen (CTA)5 and is not strongly expressed in most other normal tissues. PRAME is positively expressed in about half of uveal melanomas,6 and the majority of mucosal melanomas.7

Quality

  • United States - IVD
  • Canada - RUO
  • European Union - IVD
  • Japan - RUO

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and < 0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and format
Note: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frances Wadelin et al.
Molecular cancer, 9, 226-226 (2010-08-31)
PRAME/MAPE/OIP4 is a germinal tissue-specific gene that is also expressed at high levels in haematological malignancies and solid tumours. The physiological functions of PRAME in normal and tumour cells are unknown, although a role in the regulation of retinoic acid
Aimi Toyama et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 32(12), 1727-1733 (2019-08-04)
Mucosal melanomas are rare, and less is known about the biomarkers of this subtype in comparison to cutaneous or uveal melanomas. Preferentially expressed antigen in melanoma (PRAME) has been studied as a tool for prognostication of uveal melanomas, and immunotherapy
Cecilia Lezcano et al.
The American journal of surgical pathology, 44(4), 503-508 (2019-10-22)
The distinction of metastatic melanoma from melanocytic nevi in lymph nodes can on occasion be difficult. As diffuse immunohistochemical (IHC) PRAME (PReferentially expressed Antigen in MElanoma) expression is detected in the majority of primary and metastatic melanomas, but rarely in
Gülçin Gezgin et al.
JAMA ophthalmology, 135(6), 541-549 (2017-04-28)
Uveal melanoma (UM) is an intraocular primary malignant neoplasm that often gives rise to metastatic disease for which there are no effective therapies. A substantial proportion of UMs express the cancer-testis antigen PRAME (preferentially expressed antigen in melanoma), which can
H Ikeda et al.
Immunity, 6(2), 199-208 (1997-02-01)
Melanoma lines MEL.A and MEL.B were derived from metastases removed from patient LB33 in 1988 and 1993, respectively. The MEL.A cells express several antigens recognized by autologous cytolytic T lymphocytes (CTL) on HLA class I molecules. The MEL.B cells have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service