Skip to Content
Merck
All Photos(1)

Documents

359A-1

Sigma-Aldrich

Claudin 1 Rabbit Polyclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

Ig fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (359A-14)
vial of 0.5 mL concentrate (359A-15)
bottle of 1.0 mL predilute (359A-17)
vial of 1.0 mL concentrate (359A-16)
bottle of 7.0 mL predilute (359A-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

control

colon carcinoma, neurofibroma

shipped in

wet ice

storage temp.

2-8°C

visualization

membranous

Gene Information

human ... CLDN1(9076)

General description

The claudins are a family of over twenty proteins which are components of tight junctions. Tight junctions are specialized regions of cell-to-cell contact made up of a network of strands to act as a molecular ‘gasket′ for preventing the leakage of ions, water, etc. between cells. They are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. The claudins constitute a variable component, with specific claudins being associated with specific tissues. The immunoreactivity for anti-claudin 1 is membranous and is found in nearly all carcinomas. The staining is much stronger in the carcinoma cells than in normal tissues. Anti-claudin 1 in a panel of immunostains that includes antibodies against EMA (positive), S-100 (negative), and GLUT1 can be utilized as a robust marker in the diagnosis of perineurioma and neurofibroma. Some studies have shown anti-claudin 1 to be a specific marker for meningiomas. Therefore, anti-claudin 1 with anti-EMA, anti-S-100 protein, anti-CD34, and anti-glial fibrillary acidic protein (GFAP) may be helpful in the differentiation of meningiomas from histologic mimics.

Quality


IVD

IVD

IVD

RUO

Linkage

Claudin 1 Positive Control Slides, Product No. 359S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew L Folpe et al.
The American journal of surgical pathology, 26(12), 1620-1626 (2002-12-03)
Perineuriomas are rare benign soft tissue tumors having an immunophenotype paralleling the normal perineurial cell [S-100 protein negative and epithelial membrane antigen (EMA) positive]. Because EMA expression in perineuriomas may be focal and/or faint, there is continued interest in the
Ricardo S Macarenco et al.
Archives of pathology & laboratory medicine, 131(4), 625-636 (2007-04-12)
Perineuriomas are benign peripheral nerve sheath neoplasms composed of perineurial cells with characteristic immunohistochemical and ultrastructural features. They have been traditionally classified into two main types according to their location--intraneural and extraneural--and overlap histologically with many other tumors, which may
J Roulson et al.
Histopathology, 47(6), 551-559 (2005-12-06)
The autopsy is in decline, despite the fact that accurate mortality statistics remain essential for public health and health service planning. The falling autopsy rate combined with the Coroners Review and Human Tissue Act have contributed to this decline, and
M E F Smith et al.
Histopathology, 47(6), 575-581 (2005-12-06)
To investigate the differentiation pattern of epithelioid sarcoma in terms of perineurial and endothelial differentiation, and its relationship to that of meningioma. Nine cases of epithelioid sarcoma and five cases of meningioma were studied in an immunohistochemical analysis of 'perineurial'
Jason L Hornick et al.
The American journal of surgical pathology, 29(7), 845-858 (2005-06-17)
Perineuriomas are uncommon benign peripheral nerve sheath tumors that include soft tissue, sclerosing, and intraneural variants. Fewer than 50 soft tissue perineuriomas have been reported to date, and the clinical significance of atypical histologic features is unknown. To characterize these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service