Skip to Content
Merck
All Photos(1)

Key Documents

209A-1

Sigma-Aldrich

LH Rabbit Polyclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

Ig fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (209A-14)
vial of 0.5 mL concentrate (209A-15)
bottle of 1.0 mL predilute (209A-17)
vial of 1.0 mL concentrate (209A-16)
bottle of 7.0 mL predilute (209A-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

control

pituitary

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

General description

Anti-LH is a useful marker in classification of pituitary tumors and the study of pituitary disease. It reacts with LH-producing cells (gonadotrophs).

Quality


IVD

IVD

IVD

RUO

Linkage

LH Positive Control Slides, Product No. 209S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Sano et al.
Virchows Archiv. A, Pathological anatomy and histopathology, 417(4), 361-367 (1990-01-01)
Two pituitary adenomas removed from a 37-year-old woman and a 26-year-old woman with typical Cushing's disease were studied by light and electron microscopy, immunohistochemistry and radioimmunoassay of tissue culture media. Both patients had high plasma levels of cortisol and normal
K Saccomanno et al.
The Journal of clinical endocrinology and metabolism, 78(5), 1103-1107 (1994-05-01)
Human nonfunctioning pituitary adenomas (NFPA) may produce CG in addition to the classical glycoprotein hormones (LH, FSH, and TSH). The aim of the present study was to localize LH beta, FSH beta, TSH beta, alpha-subunit (alpha SU), CG, and its
I Felix et al.
Human pathology, 22(7), 719-721 (1991-07-01)
A 19-year-old man with blurred vision, headache, and no signs or symptoms of hormone excess was found to have a pituitary adenoma. The tumor was removed by a transfrontal approach. He had postoperative radiation therapy, but subsequently had three recurrences
J J Kovalic et al.
Journal of neuro-oncology, 16(3), 227-232 (1993-06-01)
There is general agreement that postoperative radiation therapy is beneficial for patients with subtotally resected pituitary adenomas. We have identified 41 such patients treated during a 20-year period who received postoperative irradiation for a pituitary adenoma. The usual dose was
S La Rosa et al.
Virchows Archiv : an international journal of pathology, 437(3), 264-269 (2000-10-19)
Gonadotropin-releasing hormone (GnRH), which is a well-known regulator of gonadotroph function, has recently been considered to be a paracrine factor involved in the control of somatotroph, lactotroph, and corticotroph cells. GnRH action is initiated by binding to a specific cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service