Skip to Content
Merck
All Photos(1)

Documents

20989

Sigma-Aldrich

Cesium fluoride

BioUltra, ≥99.0% (F)

Synonym(s):

NSC 84270

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CsF
CAS Number:
Molecular Weight:
151.90
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.25

product line

BioUltra

Assay

≥99.0% (F)

impurities

insoluble matter, passes filter test
≤0.005% total nitrogen (N)

loss

≤0.5% loss on drying, 110 °C

pH

6.5-7.5 (25 °C, 3 M in H2O)

mp

682 °C (lit.)

solubility

H2O: 3 M at 20 °C, clear, colorless

density

4.115 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤500 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤10 mg/kg
As: ≤0.5 mg/kg
Ba: ≤10 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤30 mg/kg
K: ≤100 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

3 M in H2O

UV absorption

λ: 260 nm Amax: 0.05
λ: 280 nm Amax: 0.05

SMILES string

[F-].[Cs+]

InChI

1S/Cs.FH/h;1H/q+1;/p-1

InChI key

XJHCXCQVJFPJIK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Preparation of building blocks for synthesis of fluoroallylic compounds
  • Synthesis of alcohols via hydrolysis of alkyl silyl ethers at neutral pH in buffered mixed organic-aqueous solutions
  • Nucleophilic fluorination of alkynyliodonium salts to form fluorovinylic compounds
  • Nucleophilic aromatic substitution (SNAr) reactions
Used in the successful synthesis of a single-crystal Dion-Jacobson phase, CsLaTa2O7, that has applications in photocatalysis and superconductivity.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Repr. 2 - STOT RE 2

Target Organs

Kidney,Adrenal gland

Supplementary Hazards

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Y Sato et al.
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 114(11), 880-887 (1994-11-01)
Benzylammonium N-alkylides were generated by reaction of N-[1-(trimethylsilyl)alkyl]benzylammonium salts with cesium fluoride in DMF or HMPA, and their rearrangement products were investigated. [2,3]Sigmatropic rearrangement of the ylides initially occurred to give isotoluene derivatives and then they were transformed into Sommelet-Hauser
N Kristianpoller et al.
Radiation protection dosimetry, 100(1-4), 207-209 (2002-10-18)
Optical and dosimetric properties of nominally pure CsGd2F7 crystals and of CsGd2F7 crystals doped with various concentrations of Pr3+ ions were investigated. Effects of X, beta and UV irradiation on these crystals were studied. Methods of optical absorption, X and
J Kypr et al.
International journal of biological macromolecules, 13(1), 9-13 (1991-02-01)
Ten DNA fragments containing self-complementary alternating sequences of adenine and thymine differing in length and the starting nucleotide were studied by c.d. spectroscopy. It was found that d(TATATATA) but not d(ATATATAT), d(TATATA), d(CTATATAG) or (dT-dA)20 isomerized into the unusual X-DNA
E D Risby et al.
Archives of general psychiatry, 48(6), 513-524 (1991-06-01)
As part of a study of the effects of lithium carbonate on neurochemical function in man, platelet and lymphocyte adenylate cyclase activity and lymphocyte beta-adrenergic receptor binding characteristics were determined before and after 2 weeks of lithium treatment in 10
E A Lesnik et al.
Journal of biomolecular structure & dynamics, 5(3), 601-614 (1987-12-01)
We have determined the 1H----3H exchange rate constants between water and C8H groups of purinic residues of alternating polynucleotides poly(dA-dT).poly(dA-dT), poly(dG-dC).poly(dG-dC) and poly(dA-dC).poly(dG-dT) as well as homopolynucleotides poly(dA).poly(dT) and poly(dG).poly(dC) in aqueous solutions with high-salt concentrations (3 M NaCl and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service