Key Documents
1509P
Millipore
BIOEAZE Tubing and Components
About This Item
UNSPSC Code:
12352202
description
Colder Quick Connect - Autoclavable
packaging
pkg of 25 pkg
General description
Female Quick Connect Cap Polycarbonate for 1/4" and 3/8" Male Quick Connect
Legal Information
BIOEAZE is a trademark of Sigma-Aldrich Co. LLC