Skip to Content
Merck
All Photos(1)

Key Documents

Y0001630

Prednicarbate for system suitability A

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Prednicarbate, (11β)-17-[(Ethoxycarbonyl)oxy]-11-hydroxy-21-(1-oxopropoxy)pregna-1,4-diene-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H36O8
CAS Number:
Molecular Weight:
488.57
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

prednicarbate

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O(C1(C2(C(C3C(C4(C(=CC(=O)C=C4)CC3)C)C(C2)O)CC1)C)C(=O)COC(=O)CC)C(=O)OCC

InChI

1S/C27H36O8/c1-5-22(31)34-15-21(30)27(35-24(32)33-6-2)12-10-19-18-8-7-16-13-17(28)9-11-25(16,3)23(18)20(29)14-26(19,27)4/h9,11,13,18-20,23,29H,5-8,10,12,14-15H2,1-4H3

InChI key

FNPXMHRZILFCKX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Prednicarbate for system suitability A EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aditya K Gupta et al.
Journal of cutaneous medicine and surgery, 8(4), 244-247 (2005-08-11)
Topical steroids have been a popular choice for treating various cutaneous disorders; however, the potential for significant local and systemic adverse events, like skin atrophy and hypothalamic-pituitary-adrenal (HPA) axis suppression, has limited their use. This article reviews the topical steroid
Kerstin Katzer et al.
European journal of dermatology : EJD, 20(1), 82-84 (2009-10-03)
The epidermal growth factor receptor (EGFR) inhibitor cetuximab is used to treat advanced stages of solid tumors. Acneiform skin eruptions, the most common side effects of cetuximab, stigmatize the patient in daily life and may lower compliance. As oral treatment
Aditya K Gupta et al.
Journal of drugs in dermatology : JDD, 3(5), 553-556 (2004-11-24)
Prednicarbate is a non-halogenated, double-ester derivative of prednisolone that has become of interest for the treatment of inflammatory skin diseases, for example atopic dermatitis. In the past, topical steroids have been a popular choice for treating these diseases; however, the
I Spika et al.
Skin pharmacology and applied skin physiology, 16(3), 143-150 (2003-04-05)
As compared to standard glucocorticoids (GC), prednicarbate (PC) is favorable in the treatment of eczema due to its high benefit/risk ratio. The remarkable anti-inflammatory effects of PC are in strong contrast to its reported low glucocorticoid receptor (GR) binding affinity.
A K Gupta et al.
Skin therapy letter, 9(10), 5-6 (2005-01-20)
Prednicarbate is a nonhalogenated corticosteroid that is used in the treatment of inflammatory skin diseases, for example atopic dermatitis. It has a favorable benefit-risk ratio, with an inflammatory action similar to that of a medium potency corticosteroid, but with a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service