Skip to Content
Merck
All Photos(3)

Key Documents

S5651

Sigma-Aldrich

Starch from potato

suitable for electrophoresis

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NB.22

biological source

potato

Quality Level

description

Hydrolyzed

form

powder

technique(s)

electrophoresis: suitable

impurities

<6% acetone

color

white to off-white

application(s)

agriculture

storage temp.

room temp

Application


  • Development and application of mathematical modeling of thymol release from environmental-responsive potato starch active packaging films.: This paper presents the development and application of mathematical models to predict thymol release from potato starch-based active packaging films. These films respond to environmental changes, making them suitable for food preservation applications (Cui et al., 2024).

  • Applications and characterization of anti-browning enzymatically modified potato starch (EPS) film associated with chitosan (CTS)/L-Cys/citric acid (CA) on fresh-cut potato slices.: This research explores the use of enzymatically modified potato starch films in combination with chitosan, L-cysteine, and citric acid to prevent browning in fresh-cut potatoes, enhancing their shelf life and quality (Zhu et al., 2024).

  • Effects of synergistic action on rheological and thermal properties of potato starch complexes co-gelatinized with caffeic acid and squash polysaccharides extracted with water and subcritical water.: This study examines the rheological and thermal properties of potato starch complexes when co-gelatinized with caffeic acid and squash polysaccharides. The findings suggest potential applications in food and biopolymer industries (Zhang et al., 2024).

Preparation Note

Hydrolyzed for electrophoresis

Analysis Note

This high quality starch is specifically tested for use in starch gel electrophoresis.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicholas Arpaia et al.
Nature, 504(7480), 451-455 (2013-11-15)
Intestinal microbes provide multicellular hosts with nutrients and confer resistance to infection. The delicate balance between pro- and anti-inflammatory mechanisms, essential for gut immune homeostasis, is affected by the composition of the commensal microbial community. Regulatory T cells (Treg cells)
Daniëlle Haenen et al.
The Journal of nutrition, 143(12), 1889-1898 (2013-10-18)
Consumption of resistant starch (RS) has been associated with various intestinal health benefits, but knowledge of its effects on global gene expression in the colon is limited. The main objective of the current study was to identify genes affected by
Shujun Wang et al.
Food & function, 4(11), 1564-1580 (2013-10-08)
Starch is the most important glycemic carbohydrate in foods. The relationship between the rate and extent of starch digestion to produce glucose for absorption into the bloodstream and risk factors for diet-related diseases is of considerable nutritional interest. Native starch
Karolien Decamps et al.
Food chemistry, 148, 235-239 (2013-11-23)
The impact of pyranose oxidase (P₂O), glucose oxidase (GO) and H₂O₂ on gluten agglomeration during wheat flour gluten-starch separation was studied. Analysis of gluten aggregate sizes in batter formed from wheat flour dough revealed that increasing levels of oxidising agents
Mark Stitt et al.
Current opinion in plant biology, 15(3), 282-292 (2012-05-01)
Many plants store part of their photosynthate as starch during the day and remobilise it to support metabolism and growth at night. Mutants unable to synthesize or degrade starch show strongly impaired growth except in long day conditions. In rapidly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service