PHR3483
Buspirone Related Compound L
certified reference material, pharmaceutical secondary standard
Synonym(s):
8-(4-Chlorobutyl)-8-azaspiro[4.5]decane-7,9-dione
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Quality Level
Agency
traceable to USP 1078824
CofA
current certificate can be downloaded
packaging
pkg of 150 mg
application(s)
pharmaceutical
storage temp.
2-8°C
SMILES string
ClCCCCN1C(=O)CC2(CCCC2)CC1=O
InChI
1S/C13H20ClNO2/c14-7-3-4-8-15-11(16)9-13(10-12(15)17)5-1-2-6-13/h1-10H2
Looking for similar products? Visit Product Comparison Guide
General description
Application
Analysis Note
Other Notes
Footnote
related product
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service