Skip to Content
Merck
All Photos(1)

Documents

PHR1475

Supelco

Disodium Inosinate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Inosine 5′-monophosphate disodium salt hydrate, 5′-IMP-Na2, 5′-Inosinic acid disodium salt hydrate, I-5′-P, IMP, Inosinic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11N4O8PNa2 · xH2O
CAS Number:
Molecular Weight:
392.17 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1222002

API family

disodium inosinate

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP([O-])([O-])=O)n2cnc3C(=O)NC=Nc23

InChI

1S/C10H13N4O8P.2Na.H2O/c15-6-4(1-21-23(18,19)20)22-10(7(6)16)14-3-13-5-8(14)11-2-12-9(5)17;;;/h2-4,6-7,10,15-16H,1H2,(H,11,12,17)(H2,18,19,20);;;1H2/q;2*+1;/p-2/t4-,6-,7-,10-;;;/m1.../s1

InChI key

KQCIGEIXDXQSGM-MSQVLRTGSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Disodium inosinate is the disodium salt of inosinic acid which belongs to the group of purine-5′-nucleotides. It is generally used as a taste and flavor enhancer in foods to provide umami taste.

Application

Disodium inosinate may be used as a pharmaceutical reference standard for the determination of the analyte in raw materials and food products by spectrophotometry, and chromatography.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Inosine monophosphate, or inosinic acid, is the ribonucleotide of hypoxanthine and the first nucleotide formed during purine synthesis.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA3023 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of purity reference material of disodium inosinate [J]
GAO YH, et al.
China Measurement and Testing, 6 (2009)
Determination of IMP and GMP in Flavor Nucleotides by HPLC [J]
LI YJ, et al.
Chinese Journal of Experimental Medical Formulae, 7 (2012)
Simultaneous UV-Spectrophotometric Determination of disodium 5?-inosinate and Disodium 5?-guanylate in tasty flavourings with calibration by the method of Principal Component Regression [J]
Shan YE
Physical Testing and Chemical Analysis Part B:Chemical Analysis, 7 (2010)
Taste perception and Integration
Flavor, 101-119 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service