Skip to Content
Merck
All Photos(1)

Documents

PHR1043

Supelco

Prednisolone

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

1,4-Pregnadiene-11β,17α,21-triol-3,20-dione, 1-Dehydrocortisol, 1-Dehydrohydrocortisone, 11β,17α,21-Trihydroxy-1,4-pregnadiene-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28O5
CAS Number:
Molecular Weight:
360.44
Beilstein:
1354103
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 464
traceable to Ph. Eur. P2700000
traceable to USP 1555005

API family

prednisolone

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

240 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

O=C1C=C[C@@]2(C)C(CC[C@]3([H])[C@]2([H])[C@@H](O)C[C@@]4(C)[C@@]3([H])CC[C@]4(O)C(CO)=O)=C1

InChI

1S/C21H28O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h5,7,9,14-16,18,22,24,26H,3-4,6,8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1

InChI key

OIGNJSKKLXVSLS-VWUMJDOOSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Prednisolone is a glucocorticoid utilized as an anti-inflammatory or an immunosuppressive drug.

Application

Prednisolone may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography techniques.
Prednisolone may be used as a standard in the separation and determination of prednisolone in serum samples of animals using radioimmunoassay.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA6903 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative determination of captopril and prednisolone in tablets by FT-Raman spectroscopy
Mazurek S and Szostak R
Journal of Pharmaceutical and Biomedical Analysis, 40(5), 1225-1230 (2006)
Development and validation of spectrophotometric and HPLC method for the simultaneous estimation of salbutamol sulphate and prednisolone in tablet dosage form
Chitlange SS, et al.
Journal of Analytical and Bioanalytical Techniques , 2(117), 2-2 (2011)
Separation of serum prednisolone and prednisolone-21-hemisuccinate by extraction and their concurrent determination by radioimmunoassay
Yanaglbashl, K, et al.
Clinical Chemistry, 26(9), 1301-1303 (1980)
Axel Heidenreich et al.
European journal of cancer (Oxford, England : 1990), 50(6), 1090-1099 (2014-02-04)
Cabazitaxel/prednisone has been shown to prolong survival versus mitoxantrone/prednisone in patients with metastatic castration-resistant prostate cancer (mCRPC) that has progressed during or after docetaxel. Subsequently, compassionate-use programmes (CUPs) and expanded-access programmes (EAPs) were established worldwide, allowing access to cabazitaxel before
J A P Spijkers-Hagelstein et al.
Leukemia, 28(4), 761-769 (2013-08-21)
Successful treatment results for MLL-rearranged Acute Lymphoblastic Leukemia (ALL) in infants remain difficult to achieve. Significantly contributing to therapy failure is poor response to glucocorticoids (GCs), like prednisone. Thus, overcoming resistance to these drugs may be a crucial step towards

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service