Skip to Content
Merck
All Photos(1)

Key Documents

P2939060

Prilocaine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

N-(2-Methylphenyl)-2-(propylamino)propanamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20N2O · HCl
CAS Number:
Molecular Weight:
256.77
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

prilocaine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].CCCNC(C)C(=O)Nc1ccccc1C

InChI

1S/C13H20N2O.ClH/c1-4-9-14-11(3)13(16)15-12-8-6-5-7-10(12)2;/h5-8,11,14H,4,9H2,1-3H3,(H,15,16);1H

InChI key

BJPJNTKRKALCPP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Prilocaine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gareth Rooke et al.
The New Zealand medical journal, 127(1407), 75-79 (2014-12-23)
This case report describes a rare side effect during a Bier's block. During local anaesthetic injection, the patient suffered a sudden onset painful petechial rash localised to the upper limb, distal to the tourniquet, without systemic effect. After deflation of
Improving the local anesthesia experience for our patients.
Gregori M Kurtzman
Dentistry today, 33(4), 120-120 (2014-05-06)
Aykut Saritas et al.
JPMA. The Journal of the Pakistan Medical Association, 64(4), 433-436 (2014-05-29)
To determine whether the addition of 8 mg dexamethasone to axillary brachial plexus block would prolong the duration of sensory and motor block in patients undergoing hand and forearm surgery. The prospective, randomised, double-blinded study was conducted at the Eskisehir
Oded Shamriz et al.
The Israel Medical Association journal : IMAJ, 16(4), 250-254 (2014-05-20)
With growing awareness of the importance of pain control in all procedures, the use of lidocaine-prilocaine cream (EMLA) for all ages is increasing. Lidocaine-prilocaine cream has been implicated as a cause of methemoglobinemia. Diagnostic clues may be oxygen-resistant cyanosis and
Susan Parekh et al.
The Cochrane database of systematic reviews, 12(12), CD009742-CD009742 (2014-12-24)
Whilst carrying out dental procedures under general anaesthesia (GA), practitioners routinely give local anaesthetics (LA) intraoperatively to children. Local anaesthetics are used to help manage postoperative pain and reduce bleeding and the physiological response to procedures. Studies of effectiveness of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service