Skip to Content
Merck
All Photos(1)

Key Documents

P2130000

Piroxicam

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-Hydroxy-2-methyl-3-(pyrid-2-yl-carbamoyl)-2H-1,2-benzothiazine 1,1-dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13N3O4S
CAS Number:
Molecular Weight:
331.35
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

piroxicam

manufacturer/tradename

EDQM

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CN1C(C(=O)Nc2ccccn2)=C(O)c3ccccc3S1(=O)=O

InChI

1S/C15H13N3O4S/c1-18-13(15(20)17-12-8-4-5-9-16-12)14(19)10-6-2-3-7-11(10)23(18,21)22/h2-9,19H,1H3,(H,16,17,20)

InChI key

QYSPLQLAKJAUJT-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Piroxicam EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Cyclooxygenase inhibitor.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - STOT RE 2 Oral

Target Organs

Gastrointestinal tract

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael A Kron et al.
Clinical and vaccine immunology : CVI, 20(2), 276-281 (2012-12-21)
The therapeutic effects of a controlled parasitic nematode infection on the course of inflammatory bowel disease (IBD) have been demonstrated in both animal and human models. However, the inability of individual well-characterized nematode proteins to recreate these beneficial effects has
Jian X Wu et al.
Journal of pharmaceutical sciences, 102(3), 904-914 (2012-12-06)
Several factors with complex interactions influence the physical stability of solid dispersions, thus highlighting the need for efficient experimental design together with robust and simple multivariate model. Design of Experiments together with ANalysis Of VAriance (ANOVA) model is one of
Xin-Yue Song et al.
Talanta, 100, 153-161 (2012-11-13)
A novel design of carbon nanotubes reinforced hollow fiber solid/liquid phase microextraction (CNTs-HF-SLPME) was developed to determine piroxicam and diclofenac in different real water samples. Functionalized multi-walled carbon nanotubes (MWCNTs) were held in the pores of hollow fiber with sol-gel
Andres Lust et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 48(1-2), 47-54 (2012-10-23)
The aim of this study was to gain understanding about the effects of different solid-state forms of a poorly water-soluble piroxicam on drug dissolution and oral bioavailability in rats. Three different solid-state forms of piroxicam were studied: anhydrate I (AH)
C R Lee et al.
Drugs, 48(6), 907-929 (1994-12-01)
Piroxicam-beta-cyclodextrin is a complex of the established nonsteroidal antiinflammatory drug (NSAID) piroxicam and an inert cyclic macromolecule, beta-cyclodextrin. In clinical trials in patients with rheumatic diseases or pain arising from other conditions, it was as effective an analgesic as standard

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service