Skip to Content
Merck
All Photos(1)

Key Documents

O1378

Supelco

Oxycodone hydrochloride

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21NO4 · HCl
CAS Number:
Molecular Weight:
351.82
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

drug control

USDEA Schedule II; Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

Cl.[H][C@@]12Oc3c(OC)ccc4C[C@H]5N(C)CC[C@@]1(c34)[C@@]5(O)CCC2=O

InChI

1S/C18H21NO4.ClH/c1-19-8-7-17-14-10-3-4-12(22-2)15(14)23-16(17)11(20)5-6-18(17,21)13(19)9-10;/h3-4,13,16,21H,5-9H2,1-2H3;1H/t13-,16+,17+,18-;/m1./s1

InChI key

MUZQPDBAOYKNLO-RKXJKUSZSA-N

Gene Information

human ... OPRM1(4988)

Looking for similar products? Visit Product Comparison Guide

General description

Oxycodone hydrochloride is an analgesic drug, which is generally used in cancer treatment since it can reduce the pain induced during the treatment.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

μ and κ opioid receptor agonist.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clinical observations on safety and efficacy of OxyContin? administered by rectal route in treating cancer related pain
Li G-C, et al.
Asian Pacific Journal of Cancer Prevention, 12, 2477-2478 (2011)
Rachel E Martin et al.
eNeuro, 8(4) (2021-07-28)
Opioid drugs are increasingly being prescribed to pregnant women. Such compounds can also bind and activate opioid receptors in the fetal brain, which could lead to long-term brain and behavioral disruptions. We hypothesized that maternal treatment with oxycodone (OXY), the
M Kokki et al.
British journal of anaesthesia, 112(1), 133-140 (2013-10-18)
Despite being increasingly used for pain management, only two studies, with controversial results, have evaluated the epidural use of oxycodone. Twenty-four women, aged 26-64 yr, undergoing elective gynaecological surgery were enrolled in this randomized, double-blinded, parallel group study. The subjects
Self-assembly of thermally responsive nanoparticles of a genetically encoded peptide polymer by drug conjugation.
Jonathan R McDaniel et al.
Angewandte Chemie (International ed. in English), 52(6), 1683-1687 (2013-01-03)
M Soledad Cepeda et al.
The journal of pain : official journal of the American Pain Society, 14(2), 158-164 (2012-12-21)
Obtaining opioids from multiple prescribers, known as doctor shopping, is 1 example of opioid abuse and diversion. The dual mechanism of action of tapentadol could make tapentadol less likely to be abused than other opioids. The aim of this retrospective

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service