Skip to Content
Merck
All Photos(1)

Key Documents

NIST1486

Bone meal

NIST® SRM® 1486

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

amorphous (powder)

packaging

pkg of 50 g

manufacturer/tradename

NIST®

technique(s)

mass spectrometry (MS): suitable
optical emission spectroscopy (OES): suitable

application(s)

food and beverages
forensics and toxicology

format

matrix material

General description

This bone meal standard reference material (SRM) contains steamed bone meal that was sieved and blended to a high degree of homogeneity. A unit of SRM 1486 consists of approximately 50 g of bone meal.

NIST1486_Cert

NIST1486_SDS

Application

This bone meal standard reference material (SRM) is intended primarily for use in evaluating analytical methods used for the determination of selected major, minor, and trace elements in bone and in the material of a similar matrix.

Other Notes

  • Details on expiry, usage, and drying procedure are available in the NIST certificate.
  • Notes on handling, storage, regulations, transportation, and disposal are provided in the SDS.
Matrix Group: Animal Feed

Preparation Note

  • Certified mass fraction values for constituent elements are determined on a dry-mass basis.
  • Reference values are non-certified values that are the best estimate of the true value.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nattida Charadram et al.
Journal of structural biology, 181(3), 207-222 (2012-12-25)
In response to microbial invasion of dentin odontoblasts secrete an altered calcified matrix termed reactionary dentin (Rd). 3D reconstruction of focused-ion-beam scanning electron microscopy (FIB-SEM) image slices revealed helical tubular structures in Rd that contrasted with regular cylindrical tubules characteristic
J K Pringle et al.
Forensic science international, 332, 111175-111175 (2022-01-14)
Hand-held, portable X-Ray fluorescence instruments (pXRF) provide a means of rapid, in-situ chemical characterisation that has considerable application as a rapid trace evidence characterisation tool in forensic geoscience. This study presents both a control test study which demonstrates optimisation of
Thomas H Darrah et al.
Metallomics : integrated biometal science, 1(6), 479-488 (2009-11-01)
We find anomalously high gadolinium (Gd) concentrations in the femoral head bones of patients exposed to chelated Gd, commonly used as a contrast agent for medical imaging. Gd is introduced in chelated form to protect patients from exposure to toxic
A review of radiologically important trace elements in human bones.
Tandon L, et al.
Applied Radiation and Isotopes, 49(8), 903-910 (1998)
Renata Rank Miranda et al.
Journal of nanobiotechnology, 18(1), 164-164 (2020-11-11)
Combination chemotherapy uses drugs that target different cancer hallmarks, resulting in synergistic or additive toxicity. This strategy enhances therapeutic efficacy as well as minimizes drug resistance and side effects. In this study, we investigated whether silver nanoparticles act as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service