Skip to Content
Merck
All Photos(1)

Key Documents

N1280000

Nortriptyline hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

3-(10,11-Dihydro-5H-dibenzo[a,d]cyclohepten-5-ylidene)-N-methyl-1-propanamine hydrochloride, Desmethylamitriptyline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21N · HCl
CAS Number:
Molecular Weight:
299.84
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

nortriptyline

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CNCC\C=C1\c2ccccc2CCc3ccccc13

InChI

1S/C19H21N.ClH/c1-20-14-6-11-19-17-9-4-2-7-15(17)12-13-16-8-3-5-10-18(16)19;/h2-5,7-11,20H,6,12-14H2,1H3;1H

InChI key

SHAYBENGXDALFF-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Nortriptyline hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rudolf Uher et al.
Depression and anxiety, 29(12), 1043-1049 (2012-08-31)
It has been suggested that clinician-rated scales and self-report questionnaires may be interchangeable in the measurement of depression severity, but it has not been tested whether clinically significant information is lost when assessment is restricted to either clinician-rated or self-report
Fabrice Gritti et al.
Journal of chromatography. A, 1282, 58-71 (2013-02-21)
The adsorption behaviors of a neutral (caffeine) and a positively charged compound (nortriptylinium) are investigated on two RPLC/hybrid stationary phases, eluted with a low ionic strength buffer (phosphate buffer, W(S)pH 2.63, I=10mM). The first phase, bridge ethylene hybrid (BEH), is
Henry P Parkman et al.
JAMA, 310(24), 2640-2649 (2013-12-26)
Gastroparesis remains a challenging syndrome to manage, with few effective treatments and a lack of rigorously controlled trials. Tricyclic antidepressants are often used to treat refractory symptoms of nausea, vomiting, and abdominal pain. Evidence from well-designed studies for this use
Aravind Penmatsa et al.
Nature, 503(7474), 85-90 (2013-09-17)
Antidepressants targeting Na(+)/Cl(-)-coupled neurotransmitter uptake define a key therapeutic strategy to treat clinical depression and neuropathic pain. However, identifying the molecular interactions that underlie the pharmacological activity of these transport inhibitors, and thus the mechanism by which the inhibitors lead
Robert A Power et al.
Journal of psychiatric research, 46(10), 1333-1338 (2012-07-10)
Most comparisons of the efficacy of antidepressants have relied on the assumption that missing data are randomly distributed. Dropout rates differ between drugs, suggesting this assumption may not hold true. This paper examines the effect of non-random dropout on a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service