Skip to Content
Merck
All Photos(1)

Key Documents

N0900000

Nitrazepam

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11N3O3
CAS Number:
Molecular Weight:
281.27
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

nitrazepam

manufacturer/tradename

EDQM

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccc2NC(=O)CN=C(c3ccccc3)c2c1

InChI

1S/C15H11N3O3/c19-14-9-16-15(10-4-2-1-3-5-10)12-8-11(18(20)21)6-7-13(12)17-14/h1-8H,9H2,(H,17,19)

InChI key

KJONHKAYOJNZEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Nitrazepam EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mihalj Posa et al.
Colloids and surfaces. B, Biointerfaces, 74(1), 84-90 (2009-07-28)
Critical micellear concentrations (CMC) were determined for two novel promoters of membrane permeability-7-monoketocholic acid (7-MKC) and 12-monoketocholic acid (12-MKC), using two non-invasive ((1)H NMR relaxation experiment and conductometry) and two invasive (spectral shift and partition coefficient of the probe molecule)
Masatomo Suetsugi et al.
Progress in neuro-psychopharmacology & biological psychiatry, 31(4), 839-847 (2007-03-03)
The first-night effect is a well-known phenomenon that is considered to result from a subject's lack of adaptation to the unfamiliar environment of a sleep laboratory and to the technical equipment used for polysomnography. The effect has been explored as
Mihalj Poša et al.
Colloids and surfaces. B, Biointerfaces, 86(2), 285-291 (2011-05-07)
The formation of mixed micelles built of 7,12-dioxolithocholic and the following hydrophobic bile acids was examined by conductometric method: cholic (C), deoxycholic (D), chenodeoxycholic (CD), 12-oxolithocholic (12-oxoL), 7-oxolithocholic (7-oxoL), ursodeoxycholic (UD) and hiodeoxycholic (HD). Interaction parameter (β) in the studied
Kuang-Chuan Wang et al.
Forensic science international, 224(1-3), 84-89 (2012-12-19)
We report determination of metabolites of popular drugs of abuse, including nimetazepam and nitrazepam, in urine by using liquid chromatography/mass spectrometry. Nimetazepam and its metabolites, 7-aminonimetazepam and nitrazepam, were extracted by solid-phase extraction using a DAU cartridge. An ammonium acetate
Katsuhiko Mizuno et al.
Drug metabolism and disposition: the biological fate of chemicals, 37(2), 345-351 (2008-11-14)
Cytochrome P450 3A4 is the predominant isoform in liver, and it metabolizes more than 50% of the clinical drugs commonly used. However, CYP3A4 is also responsible for metabolic activation of drugs, leading to liver injury. Benzodiazepines are widely used as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service