Skip to Content
Merck
All Photos(3)

Key Documents

M2772

Sigma-Aldrich

Menthol

99%

Synonym(s):

2-Isopropyl-5-methylcyclohexanol, Hexahydrothymol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O
CAS Number:
Molecular Weight:
156.27
Beilstein:
3194263
EC Number:
MDL number:
UNSPSC Code:
12352212
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

vapor pressure

0.8 mmHg ( 20 °C)

Quality Level

Assay

99%

bp

216 °C (lit.)

mp

34-36 °C (lit.)

density

0.89 g/mL at 25 °C (lit.)

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

SMILES string

CC(C)C1CCC(C)CC1O

InChI

1S/C10H20O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-11H,4-6H2,1-3H3

InChI key

NOOLISFMXDJSKH-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Menthol (C10H20O) is a terpenoid compound. It is extracted from the plants belonging to Mentha genus in the Lamiaceae family.[1]
Menthol is a cyclic monoterpene alcohol, which is extracted from essential oils of Mentha canadensis and Mentha x piperita. It possesses antimicrobial, anticancer and anti-inflammatory properties.[2] Menthol acts as a transient receptor potential cation channel (TRPM8) agonist.[3] It also exhibits analgesic, antipruritic, anesthetic and cooling effects.[2]

Application

Menthol has been used:
  • to test its antifungal susceptibility[4]
  • to determine its efficacy as an anesthetic in fish[5]
  • to activate transient receptor potential cation channel (TRPM8)[6][7]
  • as an odorant for measuring sniffing dynamics[8]

Biochem/physiol Actions

Menthol exhibits antifungal action on the membrane permeability and the cell wall consistency, this might leads to cell death or inhibition of the C. albicans filamentation. Therefore, it can be used as a potential therapeutic agent for candidiasis.[1] In addition to antifungal activity, menthol also exhibits anesthetic, antispasmodic, anti-ulcer and antiviral properties. It is considered as a safe compound for animal life. Therefore, Menthol is widely used in pharmaceuticals, cosmetics and food industries.[5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

204.8 °F

Flash Point(C)

96 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spontaneous rapid odor source localization behavior requires interhemispheric communication
Rabell JE, et al.
Current Biology, 27(10), 1542-1548 (2017)
TRP channel mediated neuronal activation and ablation in freely behaving zebrafish
Chen S, et al.
Nature Methods, 13(2), 147-147 (2015)
Efficacy of Benzocaine, Eugenol, and Menthol as Anesthetics for Freshwater Angelfish
Romaneli RD et al.
Journal of Aquatic Animal Health, 30(3), 210-216 (2018)
Inhibitory effect of menthol on expression of aspartyl proteinase 1 in fluconazole-resistant Candida albicans
Shahrzad S and Alireza K
Journal of HerbMed Pharmacology, 8(1) (2019)
TRP channels in brown and white adipogenesis from human progenitors: new therapeutic targets and the caveats associated with the common antibiotic, streptomycin
Goralczyk A, et al
Faseb Journal, 31(8), 3251-3266 (2017)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service