Skip to Content
Merck
All Photos(1)

Key Documents

L0720800

Lithocholic acid

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

3α-Hydroxy-5β-cholan-24-oic acid, 3α-Hydroxy-5β-cholanic acid, 5β-Cholan-24-oic acid-3α-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H40O3
CAS Number:
Molecular Weight:
376.57
Beilstein:
3217757
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

lithocholic acid

manufacturer/tradename

EDQM

mp

183-188 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@]12CC[C@@]3([H])[C@]4([H])CC[C@H]([C@H](C)CCC(O)=O)[C@@]4(C)CC[C@]3([H])[C@@]1(C)CC[C@@H](O)C2

InChI

1S/C24H40O3/c1-15(4-9-22(26)27)19-7-8-20-18-6-5-16-14-17(25)10-12-23(16,2)21(18)11-13-24(19,20)3/h15-21,25H,4-14H2,1-3H3,(H,26,27)/t15-,16-,17-,18+,19-,20+,21+,23+,24-/m1/s1

InChI key

SMEROWZSTRWXGI-HVATVPOCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Lithocholic acid EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benjamin L Woolbright et al.
Toxicology letters, 228(1), 56-66 (2014-04-20)
Lithocholic acid (LCA) supplementation in the diet results in intrahepatic cholestasis and bile infarcts. Previously we showed that an innate immune response is critical for cholestatic liver injury in the bile duct ligated mice. Thus, the purpose of this study
Tanmay Bera et al.
Langmuir : the ACS journal of surfaces and colloids, 29(1), 387-392 (2012-12-21)
The concentration level of bile acids is a clinical biomarker for the diagnosis of intestinal diseases because individuals suffering from intestinal diseases have a sharply increased concentration of bile acids at micromolar levels. Here, we report the detection of lithocholic
Nathalie Ménard et al.
Pharmaceutical research, 29(7), 1882-1896 (2012-03-28)
Novel surfactants made of diglutamic acid (DG) polar head linked to lithocholic, arachidonic, linoleic or stearic acids were designed for drug solubilization. Surfactants 3-D conformer and packing parameter were determined by molecular modelling and self-assembling properties by pyrene fluorescence measurements.
Yu-Guang Wang et al.
Chemico-biological interactions, 200(1), 11-20 (2012-09-18)
Licorice (LE) has been commonly used in traditional Chinese medicine (TCM) for over 4000 years to reconcile various drugs and for hepatic disorders. Glycyrrhizin is the main bioactive component isolated from LE herbs. In the present study we examined the
Simon M Vogel et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(42), 16906-16910 (2012-10-05)
The proteins MDM2 and MDM4 are key negative regulators of the tumor suppressor protein p53, which are frequently upregulated in cancer cells. They inhibit the transactivation activity of p53 by binding separately or in concert to its transactivation domain. MDM2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service