Skip to Content
Merck
All Photos(1)

Key Documents

L0551000

Levonorgestrel

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

D(−)-Norgestrel, 13β-Ethyl-17α-ethynyl-17β-hydroxygon-4-en-3-one, 17α-Ethynyl-18-homo-19-nor-testosterone, 18,19-Dinor-13β-ethyl-17β-hydroxy-4-pregnen-20-yn-3-one, 18,19-Dinor-4-pregnen-20-yn-3-one, Levonorgestrel

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28O2
CAS Number:
Molecular Weight:
312.45
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

levonorgestrel

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1CC[C@@]2(O)C#C

InChI

1S/C21H28O2/c1-3-20-11-9-17-16-8-6-15(22)13-14(16)5-7-18(17)19(20)10-12-21(20,23)4-2/h2,13,16-19,23H,3,5-12H2,1H3/t16-,17+,18+,19-,20-,21-/m0/s1

InChI key

WWYNJERNGUHSAO-XUDSTZEESA-N

Gene Information

human ... PGR(5241)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Levonorgestrel EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satu Heliövaara-Peippo et al.
American journal of obstetrics and gynecology, 209(6), 535-535 (2013-09-04)
Menorrhagia is a common problem impairing the quality of life (QOL) of many women. Both levonorgestrel-releasing intrauterine system (LNG-IUS) and hysterectomy are effective treatment modalities but no long-term comparative studies of QOL and costs exist. The objective of this study
Alice W Y Wong et al.
Obstetrics and gynecology, 121(5), 943-950 (2013-05-03)
To estimate the rate of endometrial pathology with the prophylactic use of levonorgestrel-releasing intrauterine system in women with breast cancer treated with tamoxifen. This was a randomized contro-lled trial of 129 Chinese women who attended a university hospital in Hong
Jantien Visser et al.
The Cochrane database of systematic reviews, 3(3), CD003990-CD003990 (2013-04-02)
Adequate contraceptive advice is important in both women with diabetes mellitus type 1 and type 2 to reduce the risk of maternal and infant morbidity and mortality in unplanned pregnancies. A wide variety of contraceptives are available for these women.
Anita Nelson et al.
Obstetrics and gynecology, 122(6), 1205-1213 (2013-11-19)
To evaluate the efficacy and safety of two low-dose levonorgestrel intrauterine contraceptive systems. Nulliparous and parous women aged 18-35 years with regular menstrual cycles (21-35 days) requesting contraception were randomized to 3 years of treatment with one of two levonorgestrel
H K Kroupova et al.
Aquatic toxicology (Amsterdam, Netherlands), 154, 154-162 (2014-06-04)
The aim of the present study was to investigate the effects of the synthetic progestin levonorgestrel (LNG) on the reproductive endocrine system of a teleost fish, the roach (Rutilus rutilus). Pubertal roach were exposed for 28 days in a flow-through

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service