Skip to Content
Merck
All Photos(4)

Key Documents

I7905

Sigma-Aldrich

Ibuprofen

meets USP testing specifications

Synonym(s):

α-Methyl-4-(isobutyl)phenylacetic acid, (±)-2-(4-Isobutylphenyl)propanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18O2
CAS Number:
Molecular Weight:
206.28
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
meets USP testing specifications

Quality Level

form

solid

application(s)

pharmaceutical (small molecule)

SMILES string

CC(C)Cc1ccc(cc1)C(C)C(O)=O

InChI

1S/C13H18O2/c1-9(2)8-11-4-6-12(7-5-11)10(3)13(14)15/h4-7,9-10H,8H2,1-3H3,(H,14,15)

InChI key

HEFNNWSXXWATRW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • A Case of Erythema Nodosum in a 20-Year-Old Female During the Postpartum Period.: Details a clinical case where ibuprofen was implicated in the treatment of erythema nodosum, offering insights into its therapeutic applications and associated risks (Nguyen M et al., 2024).

  • Coracoclavicular Joint Arthrosis - An Uncommon Cause of Shoulder Pain.: Investigates the use of ibuprofen in managing pain associated with rare joint disorders, contributing to broader understanding of pain management in orthopedic conditions (Graça NNJ et al., 2024).

Biochem/physiol Actions

Cyclooxygenase (COX) inhibitor that has greater activity against COX-1 than against COX-2.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J B Whitlam et al.
Clinical pharmacology and therapeutics, 29(4), 487-492 (1981-04-01)
Free and total ibuprofen levels in serum and synovial fluid (SF) were determined in one male and 14 female arthritic patients (mean age 56 yr, range 19 to 77) after 400 mg three times daily for 2 days. Free drug
R O Day et al.
Clinical pharmacology and therapeutics, 43(5), 480-487 (1988-05-01)
The simultaneous disposition of the enantiomers of ibuprofen in synovial fluid and plasma was studied in eight patients with arthritis. Concentrations of the active S-enantiomer in synovial fluid exceeded those of the R-enantiomer at all times in all patients with
Zhangjian Huang et al.
Journal of medicinal chemistry, 54(5), 1356-1364 (2011-02-02)
The carboxylic acid group of the anti-inflammatory (AI) drugs indo-methacin, (S)-naproxen and ibuprofen was covalently linked via a two-carbon ethyl spacer to a sulfohydroxamic acid moiety (CH(2)CH(2)SO(2)NHOH) to furnish a group of hybrid ester prodrugs that release nitric oxide (NO)
Carlos Velázquez et al.
Journal of medicinal chemistry, 48(12), 4061-4067 (2005-06-10)
A novel group of hybrid nitric oxide-releasing nonsteroidal antiinflammatory drugs ((*)NO-NSAIDs) possessing a 1-(pyrrolidin-1-yl)diazen-1-ium-1,2-diolate (11, 13, 15) or 1-(N,N-dimethylamino)diazen-1-ium-1,2-diolate (12, 14, 16) moiety attached via a one-carbon methylene spacer to the carboxylic acid group of the traditional NSAIDs aspirin, ibuprofen
Morshed A Chowdhury et al.
Journal of medicinal chemistry, 52(6), 1525-1529 (2009-03-20)
A novel class of 1-(4-methanesulfonylphenyl and 4-aminosulfonylphenyl)-5-[4-(1-difluoromethyl-1,2-dihydropyrid-2-one)]-3-trifluoromethyl-1H-pyrazole hybrid cyclooxygenase-2 (COX-2)/5-lipoxygenase (5-LOX) inhibitory anti-inflammatory agents was designed. Replacement of the tolyl ring present in celecoxib by the N-difluoromethyl-1,2-dihydropyrid-2-one moiety provided compounds showing dual selective COX-2/5-LOX inhibitory activities. 1-(4-Aminosulfonylphenyl)-5-[4-(1-difluoromethyl-1,2-dihydropyrid-2-one)]-3-trifluoromethyl-1H-pyrazole exhibited good anti-inflammatory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service